Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7845731-7846002 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDM6B
Entrez Gene ID
23135
Gene Name
lysine demethylase 6B
Gene Aliases
JMJD3
UniGene
Hs.223678
Gene Chromosome Location
Chr.17: 7834210-7854800 on Build GRCh38
Transcripts
NM_001080424.1 XM_005256549.3 XM_005256550.4 XM_005256551.3 XM_005256552.4 XM_006721483.3 XM_011523750.2 XM_011523752.2 XM_017024381.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6570022 c.138-5T>A p.? Chr.17: 7845867-7845867 on Build GRCh38
5828608 c.161C>A p.P54H Chr.17: 7845895-7845895 on Build GRCh38
5972468 c.147C>T p.A49A Chr.17: 7845881-7845881 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs927219615 Chr.17: 7845873-7845873 on Build GRCh38
rs540844940 Chr.17: 7845968-7845968 on Build GRCh38
rs764560394 Chr.17: 7845957-7845957 on Build GRCh38
rs774215847 Chr.17: 7845844-7845844 on Build GRCh38
rs145446870 Chr.17: 7845810-7845810 on Build GRCh38
rs372282783 Chr.17: 7845884-7845884 on Build GRCh38
rs375162097 Chr.17: 7845940-7845940 on Build GRCh38
rs757869213 Chr.17: 7845859-7845861 on Build GRCh38
rs554319751 Chr.17: 7845980-7845980 on Build GRCh38
rs373734474 Chr.17: 7845904-7845904 on Build GRCh38
rs762913388 Chr.17: 7845932-7845932 on Build GRCh38
rs763262499 Chr.17: 7845872-7845872 on Build GRCh38
rs148819146 Chr.17: 7845899-7845899 on Build GRCh38
rs759742385 Chr.17: 7845941-7845941 on Build GRCh38
rs760632279 Chr.17: 7845856-7845856 on Build GRCh38
rs976773843 Chr.17: 7845852-7845852 on Build GRCh38
rs755289949 Chr.17: 7845886-7845886 on Build GRCh38
rs961118105 Chr.17: 7845857-7845857 on Build GRCh38
rs756710688 Chr.17: 7845970-7845970 on Build GRCh38
rs751998041 Chr.17: 7845885-7845885 on Build GRCh38
rs200483669 Chr.17: 7845907-7845907 on Build GRCh38
rs971079224 Chr.17: 7845868-7845868 on Build GRCh38
rs777295095 Chr.17: 7845896-7845896 on Build GRCh38
rs747771561 Chr.17: 7845811-7845811 on Build GRCh38
rs773477615 Chr.17: 7845931-7845931 on Build GRCh38
rs901892348 Chr.17: 7845814-7845814 on Build GRCh38
rs781704985 Chr.17: 7845890-7845890 on Build GRCh38
rs927217129 Chr.17: 7845793-7845793 on Build GRCh38
rs1054274501 Chr.17: 7845795-7845795 on Build GRCh38
rs767869008 Chr.17: 7845942-7845942 on Build GRCh38
rs375982357 Chr.17: 7845840-7845840 on Build GRCh38
rs1057512699 Chr.17: 7845766-7845766 on Build GRCh38
rs201938457 Chr.17: 7845914-7845914 on Build GRCh38
rs1029688182 Chr.17: 7845831-7845831 on Build GRCh38
rs753019722 Chr.17: 7845943-7845943 on Build GRCh38
rs972515110 Chr.17: 7845862-7845862 on Build GRCh38
rs1010244116 Chr.17: 7845806-7845806 on Build GRCh38
rs897391870 Chr.17: 7845778-7845778 on Build GRCh38
rs772116368 Chr.17: 7845848-7845848 on Build GRCh38
rs759228425 Chr.17: 7845846-7845846 on Build GRCh38
rs1027130516 Chr.17: 7845805-7845805 on Build GRCh38
rs996186046 Chr.17: 7845817-7845817 on Build GRCh38
rs372387925 Chr.17: 7845915-7845915 on Build GRCh38
rs901923379 Chr.17: 7845966-7845966 on Build GRCh38
rs768670113 Chr.17: 7845919-7845919 on Build GRCh38
rs774420605 Chr.17: 7845935-7845935 on Build GRCh38
rs373907968 Chr.17: 7845827-7845827 on Build GRCh38
rs369345061 Chr.17: 7845803-7845803 on Build GRCh38
rs202081763 Chr.17: 7845894-7845894 on Build GRCh38
rs760390118 Chr.17: 7845864-7845864 on Build GRCh38
rs35274014 Chr.17: 7845933-7845934 on Build GRCh38
rs545379122 Chr.17: 7845947-7845947 on Build GRCh38
rs775557113 Chr.17: 7845851-7845851 on Build GRCh38
rs769275951 Chr.17: 7845834-7845834 on Build GRCh38
rs200419116 Chr.17: 7845843-7845843 on Build GRCh38
rs144535196 Chr.17: 7845918-7845918 on Build GRCh38
rs766687897 Chr.17: 7845934-7845934 on Build GRCh38
rs756544042 Chr.17: 7845895-7845895 on Build GRCh38
rs779737301 Chr.17: 7845861-7845864 on Build GRCh38
rs777203109 Chr.17: 7845835-7845835 on Build GRCh38
rs935854474 Chr.17: 7845794-7845794 on Build GRCh38

View Full Product Details