Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32935437-32935692 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-DMB
Entrez Gene ID
3109
Gene Name
major histocompatibility complex, class II, DM beta
Gene Aliases
D6S221E, RING7
UniGene
Hs.351279
Gene Chromosome Location
Chr.6: 32934629-32941070 on Build GRCh38
Transcripts
NM_002118.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6424195 c.626C>T p.P209L Chr.6: 32935649-32935649 on Build GRCh38
5565131 c.722G>A p.R241Q Chr.6: 32935553-32935553 on Build GRCh38
4505850 c.703C>T p.L235F Chr.6: 32935572-32935572 on Build GRCh38
6300742 c.702T>C p.S234S Chr.6: 32935573-32935573 on Build GRCh38
3349105 c.659C>A p.S220Y Chr.6: 32935616-32935616 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781548169 Chr.6: 32935575-32935575 on Build GRCh38
rs187493403 Chr.6: 32935504-32935504 on Build GRCh38
rs774065296 Chr.6: 32935628-32935628 on Build GRCh38
rs768453399 Chr.6: 32935611-32935611 on Build GRCh38
rs778237461 Chr.6: 32935553-32935553 on Build GRCh38
rs772997593 Chr.6: 32935632-32935632 on Build GRCh38
rs747488141 Chr.6: 32935559-32935559 on Build GRCh38
rs1020513349 Chr.6: 32935659-32935659 on Build GRCh38
rs922494864 Chr.6: 32935465-32935465 on Build GRCh38
rs1053462802 Chr.6: 32935464-32935464 on Build GRCh38
rs371915488 Chr.6: 32935524-32935524 on Build GRCh38
rs769554342 Chr.6: 32935488-32935488 on Build GRCh38
rs982742100 Chr.6: 32935513-32935513 on Build GRCh38
rs767546553 Chr.6: 32935631-32935631 on Build GRCh38
rs769513820 Chr.6: 32935549-32935549 on Build GRCh38
rs35339338 Chr.6: 32935635-32935636 on Build GRCh38
rs142046749 Chr.6: 32935635-32935635 on Build GRCh38
rs538995661 Chr.6: 32935461-32935461 on Build GRCh38
rs192597393 Chr.6: 32935548-32935548 on Build GRCh38
rs775109301 Chr.6: 32935492-32935492 on Build GRCh38
rs192338371 Chr.6: 32935517-32935517 on Build GRCh38
rs752932562 Chr.6: 32935547-32935547 on Build GRCh38
rs190433887 Chr.6: 32935525-32935525 on Build GRCh38
rs551307346 Chr.6: 32935640-32935640 on Build GRCh38
rs184737226 Chr.6: 32935519-32935519 on Build GRCh38
rs148615873 Chr.6: 32935568-32935568 on Build GRCh38
rs775164470 Chr.6: 32935596-32935596 on Build GRCh38
rs753394072 Chr.6: 32935636-32935636 on Build GRCh38
rs544034924 Chr.6: 32935578-32935578 on Build GRCh38
rs774294363 Chr.6: 32935512-32935512 on Build GRCh38
rs562209076 Chr.6: 32935601-32935601 on Build GRCh38
rs769273576 Chr.6: 32935585-32935585 on Build GRCh38
rs761224268 Chr.6: 32935531-32935531 on Build GRCh38
rs765601850 Chr.6: 32935544-32935544 on Build GRCh38
rs763844627 Chr.6: 32935508-32935508 on Build GRCh38
rs903426003 Chr.6: 32935595-32935595 on Build GRCh38

View Full Product Details