Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 133921136-133921397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGSF9B
Entrez Gene ID
22997
Gene Name
immunoglobulin superfamily member 9B
Gene Aliases
-
UniGene
Hs.204121
Gene Chromosome Location
Chr.11: 133908564-133956960 on Build GRCh38
Transcripts
NM_001277285.1 XM_006718795.3 XM_011542691.2 XM_011542692.2 XM_011542694.2 XM_011542695.2 XM_017017393.1 XM_017017394.1 XM_017017395.1 XM_017017396.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5473102 c.696C>T p.P232P Chr.11: 133921397-133921397 on Build GRCh38
6029345 c.800C>A p.P267H Chr.11: 133921293-133921293 on Build GRCh38
1507594 c.738C>T p.L246L Chr.11: 133921355-133921355 on Build GRCh38
281880 c.721G>A p.E241K Chr.11: 133921372-133921372 on Build GRCh38
376596 c.868G>A p.V290M Chr.11: 133921225-133921225 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs925229513 Chr.11: 133921223-133921223 on Build GRCh38
rs562625471 Chr.11: 133921373-133921373 on Build GRCh38
rs746394518 Chr.11: 133921349-133921349 on Build GRCh38
rs376960993 Chr.11: 133921331-133921331 on Build GRCh38
rs1033824782 Chr.11: 133921156-133921156 on Build GRCh38
rs760973291 Chr.11: 133921244-133921244 on Build GRCh38
rs369716530 Chr.11: 133921298-133921298 on Build GRCh38
rs757605113 Chr.11: 133921353-133921353 on Build GRCh38
rs772295879 Chr.11: 133921236-133921236 on Build GRCh38
rs1013360632 Chr.11: 133921194-133921194 on Build GRCh38
rs376800077 Chr.11: 133921292-133921292 on Build GRCh38
rs376908469 Chr.11: 133921363-133921363 on Build GRCh38
rs746456964 Chr.11: 133921231-133921231 on Build GRCh38
rs185241570 Chr.11: 133921313-133921313 on Build GRCh38
rs772827129 Chr.11: 133921330-133921330 on Build GRCh38
rs539427112 Chr.11: 133921226-133921226 on Build GRCh38
rs190045996 Chr.11: 133921316-133921316 on Build GRCh38
rs868608028 Chr.11: 133921155-133921155 on Build GRCh38
rs764694587 Chr.11: 133921317-133921317 on Build GRCh38
rs746341697 Chr.11: 133921301-133921301 on Build GRCh38
rs769018429 Chr.11: 133921315-133921315 on Build GRCh38
rs371377725 Chr.11: 133921334-133921334 on Build GRCh38
rs975455776 Chr.11: 133921249-133921249 on Build GRCh38
rs754085054 Chr.11: 133921350-133921350 on Build GRCh38
rs902318396 Chr.11: 133921374-133921374 on Build GRCh38
rs758112723 Chr.11: 133921184-133921184 on Build GRCh38
rs776267595 Chr.11: 133921252-133921252 on Build GRCh38
rs372650127 Chr.11: 133921346-133921346 on Build GRCh38
rs756408207 Chr.11: 133921195-133921195 on Build GRCh38
rs369340509 Chr.11: 133921287-133921287 on Build GRCh38
rs767488814 Chr.11: 133921348-133921348 on Build GRCh38
rs1020769230 Chr.11: 133921197-133921197 on Build GRCh38
rs779964508 Chr.11: 133921190-133921190 on Build GRCh38
rs750385595 Chr.11: 133921160-133921160 on Build GRCh38
rs933079900 Chr.11: 133921307-133921307 on Build GRCh38
rs934091585 Chr.11: 133921263-133921263 on Build GRCh38
rs766065592 Chr.11: 133921281-133921281 on Build GRCh38
rs774116125 Chr.11: 133921196-133921207 on Build GRCh38
rs772586048 Chr.11: 133921308-133921308 on Build GRCh38
rs751466329 Chr.11: 133921340-133921340 on Build GRCh38
rs778084061 Chr.11: 133921290-133921290 on Build GRCh38
rs771150072 Chr.11: 133921225-133921225 on Build GRCh38
rs775887861 Chr.11: 133921314-133921314 on Build GRCh38
rs763692997 Chr.11: 133921251-133921251 on Build GRCh38
rs759685917 Chr.11: 133921280-133921282 on Build GRCh38
rs750480606 Chr.11: 133921362-133921362 on Build GRCh38
rs181497989 Chr.11: 133921268-133921268 on Build GRCh38
rs757410424 Chr.11: 133921297-133921297 on Build GRCh38
rs940179777 Chr.11: 133921273-133921273 on Build GRCh38
rs749314712 Chr.11: 133921206-133921206 on Build GRCh38
rs979794961 Chr.11: 133921208-133921208 on Build GRCh38
rs150544201 Chr.11: 133921271-133921271 on Build GRCh38
rs779029258 Chr.11: 133921358-133921358 on Build GRCh38
rs947845439 Chr.11: 133921359-133921359 on Build GRCh38
rs775975180 Chr.11: 133921241-133921241 on Build GRCh38
rs373895799 Chr.11: 133921288-133921288 on Build GRCh38
rs778172020 Chr.11: 133921205-133921205 on Build GRCh38

View Full Product Details