Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 77614812-77615043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINGO1
Entrez Gene ID
84894
Gene Name
leucine rich repeat and Ig domain containing 1
Gene Aliases
LERN1, LRRN6A, UNQ201
UniGene
Hs.656765
Gene Chromosome Location
Chr.15: 77613024-77820900 on Build GRCh38
Transcripts
NM_001301186.1 NM_001301187.1 NM_001301189.1 NM_001301191.1 NM_001301192.1 NM_001301194.1 NM_001301195.1 NM_001301197.1 NM_001301198.1 NM_001301199.1 NM_001301200.1 NM_032808.6 XM_011522118.2 XM_017022682.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
965264 c.931C>T p.L311F Chr.15: 77614976-77614976 on Build GRCh38
4057098 c.1029T>C p.S343S Chr.15: 77614878-77614878 on Build GRCh38
965260 c.1012C>T p.R338C Chr.15: 77614895-77614895 on Build GRCh38
965262 c.998G>A p.G333D Chr.15: 77614909-77614909 on Build GRCh38
701763 c.893C>T p.P298L Chr.15: 77615014-77615014 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758132909 Chr.15: 77614913-77614913 on Build GRCh38
rs745918219 Chr.15: 77614892-77614892 on Build GRCh38
rs763006902 Chr.15: 77615012-77615012 on Build GRCh38
rs758164181 Chr.15: 77614865-77614865 on Build GRCh38
rs781478906 Chr.15: 77614890-77614890 on Build GRCh38
rs753336987 Chr.15: 77614837-77614837 on Build GRCh38
rs745975783 Chr.15: 77614926-77614926 on Build GRCh38
rs748432694 Chr.15: 77614953-77614953 on Build GRCh38
rs781531990 Chr.15: 77614925-77614925 on Build GRCh38
rs778795680 Chr.15: 77614854-77614854 on Build GRCh38
rs774831720 Chr.15: 77614946-77614946 on Build GRCh38
rs770463478 Chr.15: 77614893-77614893 on Build GRCh38
rs759061384 Chr.15: 77614973-77614973 on Build GRCh38
rs34904447 Chr.15: 77615022-77615022 on Build GRCh38
rs895897032 Chr.15: 77614991-77614991 on Build GRCh38
rs199976207 Chr.15: 77614934-77614934 on Build GRCh38
rs764660666 Chr.15: 77614977-77614977 on Build GRCh38
rs776220853 Chr.15: 77614968-77614968 on Build GRCh38
rs764018651 Chr.15: 77614914-77614914 on Build GRCh38
rs752301504 Chr.15: 77614933-77614933 on Build GRCh38
rs533195161 Chr.15: 77615009-77615009 on Build GRCh38
rs187712243 Chr.15: 77614962-77614962 on Build GRCh38
rs752469221 Chr.15: 77614859-77614859 on Build GRCh38
rs374002347 Chr.15: 77614833-77614833 on Build GRCh38
rs751300730 Chr.15: 77614918-77614918 on Build GRCh38
rs769035783 Chr.15: 77614944-77614944 on Build GRCh38
rs368266948 Chr.15: 77614911-77614911 on Build GRCh38
rs776098870 Chr.15: 77614894-77614894 on Build GRCh38
rs375759643 Chr.15: 77614899-77614899 on Build GRCh38
rs772742302 Chr.15: 77614898-77614898 on Build GRCh38
rs540796339 Chr.15: 77614877-77614877 on Build GRCh38
rs933474359 Chr.15: 77614864-77614864 on Build GRCh38
rs763246298 Chr.15: 77614895-77614895 on Build GRCh38
rs771055276 Chr.15: 77614883-77614883 on Build GRCh38
rs765997120 Chr.15: 77614900-77614900 on Build GRCh38
rs550891976 Chr.15: 77614935-77614935 on Build GRCh38
rs754960167 Chr.15: 77614839-77614839 on Build GRCh38
rs993347507 Chr.15: 77614948-77614948 on Build GRCh38
rs372907046 Chr.15: 77614912-77614912 on Build GRCh38
rs776160711 Chr.15: 77614910-77614910 on Build GRCh38
rs757600962 Chr.15: 77614921-77614921 on Build GRCh38
rs760113794 Chr.15: 77614836-77614836 on Build GRCh38
rs367921905 Chr.15: 77614881-77614881 on Build GRCh38

View Full Product Details