Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 70529820-70530089 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PALD1
Entrez Gene ID
27143
Gene Name
phosphatase domain containing, paladin 1
Gene Aliases
KIAA1274, PALD
UniGene
Hs.202351
Gene Chromosome Location
Chr.10: 70458542-70568450 on Build GRCh38
Transcripts
NM_014431.2 XM_011539637.2 XM_011539638.2 XM_017016072.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1702593 c.406G>A p.G136R Chr.10: 70530006-70530006 on Build GRCh38
5554632 c.425G>T p.G142V Chr.10: 70530025-70530025 on Build GRCh38
6436882 c.433C>T p.R145W Chr.10: 70530033-70530033 on Build GRCh38
3686980 c.293G>A p.R98H Chr.10: 70529893-70529893 on Build GRCh38
1348837 c.460G>A p.G154R Chr.10: 70530060-70530060 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747992510 Chr.10: 70529912-70529912 on Build GRCh38
rs764470628 Chr.10: 70529942-70529942 on Build GRCh38
rs765916639 Chr.10: 70529979-70529979 on Build GRCh38
rs765648109 Chr.10: 70530054-70530054 on Build GRCh38
rs766850114 Chr.10: 70530062-70530062 on Build GRCh38
rs757827719 Chr.10: 70529845-70529845 on Build GRCh38
rs141285632 Chr.10: 70530059-70530059 on Build GRCh38
rs766680339 Chr.10: 70529875-70529878 on Build GRCh38
rs769693757 Chr.10: 70529913-70529913 on Build GRCh38
rs751094458 Chr.10: 70529983-70529983 on Build GRCh38
rs779522965 Chr.10: 70529893-70529893 on Build GRCh38
rs775627617 Chr.10: 70530016-70530016 on Build GRCh38
rs770844377 Chr.10: 70530014-70530014 on Build GRCh38
rs143550137 Chr.10: 70530060-70530060 on Build GRCh38
rs2275059 Chr.10: 70529945-70529945 on Build GRCh38
rs765622269 Chr.10: 70529957-70529957 on Build GRCh38
rs768968808 Chr.10: 70530026-70530026 on Build GRCh38
rs756332383 Chr.10: 70529870-70529871 on Build GRCh38
rs777026032 Chr.10: 70530034-70530034 on Build GRCh38
rs759490074 Chr.10: 70529925-70529925 on Build GRCh38
rs372433744 Chr.10: 70529915-70529915 on Build GRCh38
rs897917963 Chr.10: 70530064-70530064 on Build GRCh38
rs202089641 Chr.10: 70529874-70529874 on Build GRCh38
rs747904062 Chr.10: 70529992-70529992 on Build GRCh38
rs373966854 Chr.10: 70529892-70529892 on Build GRCh38
rs754165073 Chr.10: 70529879-70529880 on Build GRCh38
rs559499747 Chr.10: 70530033-70530033 on Build GRCh38
rs757787452 Chr.10: 70529970-70529970 on Build GRCh38
rs754322556 Chr.10: 70529844-70529844 on Build GRCh38
rs933928475 Chr.10: 70530006-70530006 on Build GRCh38
rs267602562 Chr.10: 70530051-70530051 on Build GRCh38
rs750960787 Chr.10: 70529958-70529958 on Build GRCh38
rs144883248 Chr.10: 70529943-70529943 on Build GRCh38
rs201059316 Chr.10: 70530036-70530036 on Build GRCh38
rs780867492 Chr.10: 70529986-70529986 on Build GRCh38
rs773237217 Chr.10: 70529921-70529921 on Build GRCh38
rs370351884 Chr.10: 70529877-70529877 on Build GRCh38
rs777517892 Chr.10: 70530000-70530000 on Build GRCh38
rs368428125 Chr.10: 70529908-70529908 on Build GRCh38
rs981025179 Chr.10: 70529960-70529960 on Build GRCh38
rs755929135 Chr.10: 70529999-70529999 on Build GRCh38
rs926866425 Chr.10: 70529987-70529987 on Build GRCh38
rs149888345 Chr.10: 70529931-70529931 on Build GRCh38
rs2275060 Chr.10: 70530022-70530022 on Build GRCh38
rs749255806 Chr.10: 70530012-70530012 on Build GRCh38
rs754609623 Chr.10: 70529985-70529985 on Build GRCh38
rs760977206 Chr.10: 70529940-70529940 on Build GRCh38
rs749869688 Chr.10: 70529879-70529879 on Build GRCh38
rs746686132 Chr.10: 70529907-70529907 on Build GRCh38
rs755274089 Chr.10: 70529945-70529945 on Build GRCh38
rs759057667 Chr.10: 70530018-70530018 on Build GRCh38
rs775511613 Chr.10: 70529933-70529933 on Build GRCh38

View Full Product Details