Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136854579-136854836 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAMDC4
Entrez Gene ID
158056
Gene Name
MAM domain containing 4
Gene Aliases
AEGP
UniGene
Hs.376780
Gene Chromosome Location
Chr.9: 136852367-136860799 on Build GRCh38
Transcripts
NM_206920.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4422226 c.858G>C p.W286C Chr.9: 136854600-136854600 on Build GRCh38
6889380 c.934+1G>A p.? Chr.9: 136854677-136854677 on Build GRCh38
6889379 c.934+1G>A p.? Chr.9: 136854677-136854677 on Build GRCh38
3763798 c.873C>T p.R291R Chr.9: 136854615-136854615 on Build GRCh38
1214288 c.863G>A p.R288Q Chr.9: 136854605-136854605 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373389983 Chr.9: 136854765-136854765 on Build GRCh38
rs763573731 Chr.9: 136854757-136854757 on Build GRCh38
rs142114032 Chr.9: 136854616-136854616 on Build GRCh38
rs758279812 Chr.9: 136854722-136854722 on Build GRCh38
rs772381683 Chr.9: 136854806-136854806 on Build GRCh38
rs769996411 Chr.9: 136854647-136854647 on Build GRCh38
rs894934854 Chr.9: 136854646-136854646 on Build GRCh38
rs376890413 Chr.9: 136854725-136854725 on Build GRCh38
rs557616454 Chr.9: 136854649-136854649 on Build GRCh38
rs574605467 Chr.9: 136854661-136854661 on Build GRCh38
rs745544901 Chr.9: 136854677-136854678 on Build GRCh38
rs377080167 Chr.9: 136854756-136854756 on Build GRCh38
rs956680752 Chr.9: 136854656-136854656 on Build GRCh38
rs371688887 Chr.9: 136854696-136854696 on Build GRCh38
rs764396242 Chr.9: 136854604-136854604 on Build GRCh38
rs898940217 Chr.9: 136854600-136854600 on Build GRCh38
rs929866100 Chr.9: 136854801-136854801 on Build GRCh38
rs569550394 Chr.9: 136854605-136854605 on Build GRCh38
rs1018361678 Chr.9: 136854714-136854714 on Build GRCh38
rs758224590 Chr.9: 136854769-136854769 on Build GRCh38
rs144788206 Chr.9: 136854631-136854631 on Build GRCh38
rs767875162 Chr.9: 136854755-136854755 on Build GRCh38
rs757754981 Chr.9: 136854614-136854614 on Build GRCh38
rs141677206 Chr.9: 136854777-136854777 on Build GRCh38
rs769945203 Chr.9: 136854736-136854736 on Build GRCh38
rs769513664 Chr.9: 136854715-136854715 on Build GRCh38
rs1053559363 Chr.9: 136854642-136854642 on Build GRCh38
rs747948200 Chr.9: 136854720-136854720 on Build GRCh38
rs774766391 Chr.9: 136854746-136854746 on Build GRCh38
rs753503691 Chr.9: 136854709-136854709 on Build GRCh38
rs762305620 Chr.9: 136854668-136854668 on Build GRCh38
rs781670045 Chr.9: 136854662-136854662 on Build GRCh38
rs372819464 Chr.9: 136854715-136854715 on Build GRCh38
rs200633115 Chr.9: 136854650-136854650 on Build GRCh38
rs537044806 Chr.9: 136854672-136854672 on Build GRCh38
rs1045220189 Chr.9: 136854798-136854798 on Build GRCh38
rs377443370 Chr.9: 136854770-136854770 on Build GRCh38
rs7869864 Chr.9: 136854615-136854615 on Build GRCh38
rs977685434 Chr.9: 136854747-136854747 on Build GRCh38
rs374166053 Chr.9: 136854743-136854743 on Build GRCh38
rs375923464 Chr.9: 136854632-136854632 on Build GRCh38
rs1046195492 Chr.9: 136854778-136854778 on Build GRCh38
rs749388123 Chr.9: 136854784-136854784 on Build GRCh38
rs748900951 Chr.9: 136854622-136854622 on Build GRCh38
rs201073467 Chr.9: 136854738-136854738 on Build GRCh38
rs746920500 Chr.9: 136854773-136854773 on Build GRCh38
rs147996833 Chr.9: 136854662-136854662 on Build GRCh38
rs780178024 Chr.9: 136854780-136854780 on Build GRCh38
rs370273159 Chr.9: 136854785-136854785 on Build GRCh38
rs202122087 Chr.9: 136854729-136854729 on Build GRCh38
rs774619653 Chr.9: 136854666-136854666 on Build GRCh38
rs553215710 Chr.9: 136854760-136854760 on Build GRCh38
rs762111376 Chr.9: 136854748-136854748 on Build GRCh38
rs748405373 Chr.9: 136854805-136854805 on Build GRCh38
rs749495892 Chr.9: 136854739-136854739 on Build GRCh38
rs776835081 Chr.9: 136854636-136854636 on Build GRCh38
rs759630034 Chr.9: 136854640-136854640 on Build GRCh38
rs771313012 Chr.9: 136854779-136854779 on Build GRCh38
rs139450438 Chr.9: 136854817-136854817 on Build GRCh38
rs150548116 Chr.9: 136854803-136854803 on Build GRCh38
rs754643631 Chr.9: 136854761-136854761 on Build GRCh38
rs370177629 Chr.9: 136854758-136854758 on Build GRCh38
rs772200627 Chr.9: 136854633-136854633 on Build GRCh38
rs752401048 Chr.9: 136854767-136854767 on Build GRCh38
rs557276518 Chr.9: 136854697-136854697 on Build GRCh38
rs778762173 Chr.9: 136854716-136854716 on Build GRCh38

View Full Product Details