Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47267874-47268099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NR1H3
Entrez Gene ID
10062
Gene Name
nuclear receptor subfamily 1 group H member 3
Gene Aliases
LXR-a, LXRA, RLD-1
UniGene
Hs.438863
Gene Chromosome Location
Chr.11: 47248300-47269033 on Build GRCh38
Transcripts
NM_001130101.2 NM_001130102.2 NM_001251934.1 NM_001251935.1 NM_005693.3 XM_005252705.1 XM_005252706.1 XM_005252707.4 XM_005252709.1 XM_005252710.1 XM_005252713.3 XM_005252715.2 XM_005252716.3 XM_005252718.3 XM_006718112.1 XM_006718113.1 XM_006718115.1 XM_006718116.1 XM_011519805.2 XM_011519806.1 XM_011519807.1 XM_011519808.2 XM_017017056.1 XM_017017057.1 XM_017017058.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1135187 c.1040A>C p.N347T Chr.11: 47267964-47267964 on Build GRCh38
6412130 c.1061C>T p.A354V Chr.11: 47267985-47267985 on Build GRCh38
7092992 c.1005C>A p.F335L Chr.11: 47267929-47267929 on Build GRCh38
3666332 c.1015A>G p.I339V Chr.11: 47267939-47267939 on Build GRCh38
6889614 c.1000G>C p.E334Q Chr.11: 47267924-47267924 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761943794 Chr.11: 47268059-47268059 on Build GRCh38
rs765756601 Chr.11: 47267960-47267960 on Build GRCh38
rs138838833 Chr.11: 47267933-47267933 on Build GRCh38
rs371684575 Chr.11: 47268060-47268063 on Build GRCh38
rs758560008 Chr.11: 47267945-47267945 on Build GRCh38
rs144558044 Chr.11: 47268019-47268019 on Build GRCh38
rs760817324 Chr.11: 47267940-47267940 on Build GRCh38
rs755627128 Chr.11: 47267995-47267995 on Build GRCh38
rs777611725 Chr.11: 47267904-47267904 on Build GRCh38
rs763312945 Chr.11: 47268071-47268071 on Build GRCh38
rs776771553 Chr.11: 47268056-47268056 on Build GRCh38
rs763373811 Chr.11: 47267975-47267975 on Build GRCh38
rs770856544 Chr.11: 47268026-47268026 on Build GRCh38
rs375563752 Chr.11: 47267958-47267958 on Build GRCh38
rs766784809 Chr.11: 47267979-47267979 on Build GRCh38
rs763498622 Chr.11: 47268036-47268037 on Build GRCh38
rs201824260 Chr.11: 47268041-47268041 on Build GRCh38
rs764412620 Chr.11: 47268044-47268045 on Build GRCh38
rs749086742 Chr.11: 47267915-47267915 on Build GRCh38
rs751933114 Chr.11: 47268075-47268075 on Build GRCh38
rs751987916 Chr.11: 47267985-47267985 on Build GRCh38
rs775684021 Chr.11: 47267935-47267935 on Build GRCh38
rs149370955 Chr.11: 47267986-47267986 on Build GRCh38
rs368572277 Chr.11: 47267998-47267998 on Build GRCh38
rs368201770 Chr.11: 47268008-47268008 on Build GRCh38
rs750953460 Chr.11: 47267969-47267969 on Build GRCh38
rs767016801 Chr.11: 47268073-47268073 on Build GRCh38
rs955980179 Chr.11: 47268025-47268025 on Build GRCh38
rs769835628 Chr.11: 47268065-47268065 on Build GRCh38
rs988707716 Chr.11: 47268036-47268036 on Build GRCh38
rs753207033 Chr.11: 47267978-47267980 on Build GRCh38
rs760019068 Chr.11: 47268077-47268077 on Build GRCh38
rs765777154 Chr.11: 47267927-47267927 on Build GRCh38
rs545851308 Chr.11: 47268043-47268043 on Build GRCh38
rs370616663 Chr.11: 47267956-47267956 on Build GRCh38
rs778618057 Chr.11: 47268029-47268029 on Build GRCh38
rs768822049 Chr.11: 47268045-47268045 on Build GRCh38

View Full Product Details