Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49172793-49173044 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHDC8B
Entrez Gene ID
200942
Gene Name
kelch domain containing 8B
Gene Aliases
CHL
UniGene
Hs.13781
Gene Chromosome Location
Chr.3: 49171120-49176487 on Build GRCh38
Transcripts
NM_173546.2 XM_005264938.2 XM_005264940.4 XM_006713015.2 XM_006713016.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4494808 c.43C>T p.P15S Chr.3: 49172812-49172812 on Build GRCh38
5355382 c.104T>C p.V35A Chr.3: 49172873-49172873 on Build GRCh38
359799 c.48C>A p.P16P Chr.3: 49172817-49172817 on Build GRCh38
6874148 c.111G>T p.G37G Chr.3: 49172880-49172880 on Build GRCh38
6874147 c.108G>T p.L36F Chr.3: 49172877-49172877 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs569675399 Chr.3: 49172991-49172991 on Build GRCh38
rs765576569 Chr.3: 49172936-49172936 on Build GRCh38
rs373852462 Chr.3: 49172891-49172891 on Build GRCh38
rs749095409 Chr.3: 49172996-49172996 on Build GRCh38
rs778852918 Chr.3: 49173006-49173006 on Build GRCh38
rs529664748 Chr.3: 49172929-49172929 on Build GRCh38
rs758765503 Chr.3: 49172940-49172940 on Build GRCh38
rs370010051 Chr.3: 49172866-49172866 on Build GRCh38
rs756357057 Chr.3: 49172990-49172990 on Build GRCh38
rs747102634 Chr.3: 49172837-49172837 on Build GRCh38
rs541739860 Chr.3: 49172856-49172856 on Build GRCh38
rs149050663 Chr.3: 49172960-49172960 on Build GRCh38
rs772980133 Chr.3: 49172862-49172862 on Build GRCh38
rs559428069 Chr.3: 49172877-49172877 on Build GRCh38
rs369834697 Chr.3: 49172974-49172974 on Build GRCh38
rs374978976 Chr.3: 49172833-49172833 on Build GRCh38
rs760349783 Chr.3: 49172934-49172934 on Build GRCh38
rs745784716 Chr.3: 49173017-49173017 on Build GRCh38
rs751013548 Chr.3: 49172815-49172815 on Build GRCh38
rs879081295 Chr.3: 49172825-49172825 on Build GRCh38
rs548119617 Chr.3: 49172975-49172975 on Build GRCh38
rs750808403 Chr.3: 49172938-49172938 on Build GRCh38
rs775048628 Chr.3: 49173020-49173020 on Build GRCh38
rs770662379 Chr.3: 49173003-49173003 on Build GRCh38
rs767018829 Chr.3: 49172951-49172951 on Build GRCh38
rs748374314 Chr.3: 49172854-49172854 on Build GRCh38
rs775044801 Chr.3: 49172933-49172933 on Build GRCh38
rs916529622 Chr.3: 49172821-49172821 on Build GRCh38
rs755041339 Chr.3: 49172850-49172850 on Build GRCh38
rs530596191 Chr.3: 49173018-49173018 on Build GRCh38
rs996160256 Chr.3: 49172994-49172994 on Build GRCh38
rs1054946407 Chr.3: 49172813-49172813 on Build GRCh38
rs1051136165 Chr.3: 49172922-49172922 on Build GRCh38
rs756642170 Chr.3: 49172978-49172978 on Build GRCh38
rs902512320 Chr.3: 49172890-49172890 on Build GRCh38
rs752076206 Chr.3: 49172953-49172953 on Build GRCh38
rs758956586 Chr.3: 49172831-49172831 on Build GRCh38
rs758990859 Chr.3: 49172893-49172893 on Build GRCh38
rs368617575 Chr.3: 49172969-49172969 on Build GRCh38
rs755054855 Chr.3: 49172955-49172955 on Build GRCh38
rs1043772443 Chr.3: 49172860-49172860 on Build GRCh38
rs946616957 Chr.3: 49172830-49172830 on Build GRCh38
rs749934042 Chr.3: 49172891-49172891 on Build GRCh38
rs376897933 Chr.3: 49172957-49172957 on Build GRCh38
rs999496418 Chr.3: 49172896-49172896 on Build GRCh38
rs781302926 Chr.3: 49172851-49172851 on Build GRCh38

View Full Product Details