Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43675383-43675652 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRPS18A
Entrez Gene ID
55168
Gene Name
mitochondrial ribosomal protein S18A
Gene Aliases
HumanS18b, MRP-S18-3, MRPS18-3, S18bmt
UniGene
Hs.520149
Gene Chromosome Location
Chr.6: 43671197-43687812 on Build GRCh38
Transcripts
NM_001193343.1 NM_018135.3 XM_006715134.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4356018 c.256G>A p.V86I Chr.6: 43675614-43675614 on Build GRCh38
6276731 c.325C>A p.Q109K Chr.6: 43675545-43675545 on Build GRCh38
6348792 c.324C>A p.C108* Chr.6: 43675546-43675546 on Build GRCh38
6715463 c.316G>T p.G106C Chr.6: 43675554-43675554 on Build GRCh38
7128703 c.449C>T p.S150L Chr.6: 43675421-43675421 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368216569 Chr.6: 43675451-43675451 on Build GRCh38
rs199768351 Chr.6: 43675618-43675618 on Build GRCh38
rs749351308 Chr.6: 43675513-43675513 on Build GRCh38
rs569406382 Chr.6: 43675532-43675532 on Build GRCh38
rs759043524 Chr.6: 43675462-43675462 on Build GRCh38
rs746749172 Chr.6: 43675524-43675524 on Build GRCh38
rs560952992 Chr.6: 43675594-43675594 on Build GRCh38
rs768508712 Chr.6: 43675426-43675426 on Build GRCh38
rs780284141 Chr.6: 43675502-43675502 on Build GRCh38
rs142607556 Chr.6: 43675566-43675566 on Build GRCh38
rs750821263 Chr.6: 43675591-43675591 on Build GRCh38
rs750698155 Chr.6: 43675495-43675495 on Build GRCh38
rs749288566 Chr.6: 43675419-43675419 on Build GRCh38
rs1033916685 Chr.6: 43675549-43675549 on Build GRCh38
rs375266063 Chr.6: 43675519-43675519 on Build GRCh38
rs563206132 Chr.6: 43675553-43675553 on Build GRCh38
rs148462467 Chr.6: 43675559-43675559 on Build GRCh38
rs528228716 Chr.6: 43675614-43675614 on Build GRCh38
rs773064509 Chr.6: 43675438-43675438 on Build GRCh38
rs779120003 Chr.6: 43675622-43675622 on Build GRCh38
rs1011663993 Chr.6: 43675499-43675499 on Build GRCh38
rs757688407 Chr.6: 43675483-43675483 on Build GRCh38
rs866864847 Chr.6: 43675619-43675619 on Build GRCh38
rs753252004 Chr.6: 43675460-43675460 on Build GRCh38
rs781527101 Chr.6: 43675489-43675489 on Build GRCh38
rs761711992 Chr.6: 43675431-43675431 on Build GRCh38
rs144606291 Chr.6: 43675534-43675534 on Build GRCh38
rs201061028 Chr.6: 43675417-43675417 on Build GRCh38
rs772047306 Chr.6: 43675525-43675525 on Build GRCh38
rs777299772 Chr.6: 43675409-43675409 on Build GRCh38
rs773115392 Chr.6: 43675533-43675533 on Build GRCh38
rs371801011 Chr.6: 43675628-43675628 on Build GRCh38
rs769978383 Chr.6: 43675415-43675415 on Build GRCh38
rs766526095 Chr.6: 43675605-43675605 on Build GRCh38
rs866541092 Chr.6: 43675505-43675505 on Build GRCh38
rs751999346 Chr.6: 43675476-43675476 on Build GRCh38
rs771924805 Chr.6: 43675437-43675437 on Build GRCh38
rs774383395 Chr.6: 43675430-43675430 on Build GRCh38
rs371926018 Chr.6: 43675470-43675470 on Build GRCh38
rs768724677 Chr.6: 43675518-43675518 on Build GRCh38
rs767909772 Chr.6: 43675570-43675570 on Build GRCh38
rs756259629 Chr.6: 43675500-43675500 on Build GRCh38
rs542654147 Chr.6: 43675565-43675565 on Build GRCh38
rs1050451966 Chr.6: 43675446-43675446 on Build GRCh38

View Full Product Details