Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 19354434-19354689 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDHA1
Entrez Gene ID
5160
Gene Name
pyruvate dehydrogenase (lipoamide) alpha 1
Gene Aliases
PDHA, PDHAD, PDHCE1A, PHE1A
UniGene
Hs.530331
Gene Chromosome Location
Chr.X: 19343893-19361707 on Build GRCh38
Transcripts
NM_000284.3 NM_001173454.1 NM_001173455.1 NM_001173456.1 XM_017029574.1

SNP Detail

SNP ID Chromosome Location
rs770603511 Chr.X: 19354551-19354551 on Build GRCh38
rs765099790 Chr.X: 19354470-19354470 on Build GRCh38
rs752131202 Chr.X: 19354642-19354642 on Build GRCh38
rs1036317049 Chr.X: 19354630-19354630 on Build GRCh38
rs780856898 Chr.X: 19354472-19354472 on Build GRCh38
rs769308417 Chr.X: 19354502-19354502 on Build GRCh38
rs777678490 Chr.X: 19354530-19354530 on Build GRCh38
rs6633240 Chr.X: 19354490-19354490 on Build GRCh38
rs747591823 Chr.X: 19354479-19354479 on Build GRCh38
rs774254122 Chr.X: 19354605-19354605 on Build GRCh38
rs759269353 Chr.X: 19354607-19354607 on Build GRCh38
rs1051608554 Chr.X: 19354597-19354597 on Build GRCh38
rs796247061 Chr.X: 19354656-19354658 on Build GRCh38
rs994851714 Chr.X: 19354653-19354653 on Build GRCh38
rs749319044 Chr.X: 19354544-19354544 on Build GRCh38
rs921680836 Chr.X: 19354497-19354497 on Build GRCh38
rs368105884 Chr.X: 19354609-19354609 on Build GRCh38
rs1014621101 Chr.X: 19354477-19354477 on Build GRCh38
rs370506109 Chr.X: 19354626-19354626 on Build GRCh38
rs374502252 Chr.X: 19354628-19354628 on Build GRCh38
rs776993593 Chr.X: 19354616-19354616 on Build GRCh38

View Full Product Details