Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48757983-48758252 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF21
Entrez Gene ID
26291
Gene Name
fibroblast growth factor 21
Gene Aliases
-
UniGene
Hs.283015
Gene Chromosome Location
Chr.19: 48755559-48758330 on Build GRCh38
Transcripts
NM_019113.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3536660 c.462G>A p.R154R Chr.19: 48758052-48758052 on Build GRCh38
3536661 c.524C>T p.P175L Chr.19: 48758114-48758114 on Build GRCh38
4932050 c.456A>T p.P152P Chr.19: 48758046-48758046 on Build GRCh38
5114820 c.429G>A p.P143P Chr.19: 48758019-48758019 on Build GRCh38
365740 c.524C>A p.P175Q Chr.19: 48758114-48758114 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768888677 Chr.19: 48758064-48758064 on Build GRCh38
rs745977503 Chr.19: 48758060-48758060 on Build GRCh38
rs748313785 Chr.19: 48758100-48758101 on Build GRCh38
rs745406911 Chr.19: 48758186-48758186 on Build GRCh38
rs559156552 Chr.19: 48758098-48758098 on Build GRCh38
rs766337664 Chr.19: 48758148-48758148 on Build GRCh38
rs739320 Chr.19: 48758111-48758111 on Build GRCh38
rs779105985 Chr.19: 48758051-48758051 on Build GRCh38
rs867721982 Chr.19: 48758022-48758022 on Build GRCh38
rs933317607 Chr.19: 48758174-48758174 on Build GRCh38
rs112120725 Chr.19: 48758050-48758050 on Build GRCh38
rs3745710 Chr.19: 48758010-48758010 on Build GRCh38
rs778172084 Chr.19: 48758171-48758171 on Build GRCh38
rs754736990 Chr.19: 48758029-48758029 on Build GRCh38
rs769374090 Chr.19: 48758113-48758113 on Build GRCh38
rs751306574 Chr.19: 48758023-48758023 on Build GRCh38
rs773698761 Chr.19: 48758178-48758178 on Build GRCh38
rs964395738 Chr.19: 48758193-48758193 on Build GRCh38
rs759579781 Chr.19: 48758081-48758081 on Build GRCh38
rs758694383 Chr.19: 48758146-48758146 on Build GRCh38
rs41308776 Chr.19: 48758011-48758011 on Build GRCh38
rs371046094 Chr.19: 48758054-48758054 on Build GRCh38
rs760562529 Chr.19: 48758136-48758136 on Build GRCh38
rs200685930 Chr.19: 48758125-48758125 on Build GRCh38
rs201833005 Chr.19: 48758212-48758212 on Build GRCh38
rs868452579 Chr.19: 48758137-48758137 on Build GRCh38
rs755387755 Chr.19: 48758156-48758156 on Build GRCh38
rs777100377 Chr.19: 48758065-48758065 on Build GRCh38
rs775377031 Chr.19: 48758135-48758135 on Build GRCh38
rs781670890 Chr.19: 48758162-48758162 on Build GRCh38
rs182407076 Chr.19: 48758124-48758124 on Build GRCh38
rs202104197 Chr.19: 48758198-48758198 on Build GRCh38
rs375785872 Chr.19: 48758149-48758149 on Build GRCh38
rs540715300 Chr.19: 48758074-48758074 on Build GRCh38
rs885662 Chr.19: 48758106-48758106 on Build GRCh38
rs151216283 Chr.19: 48758163-48758163 on Build GRCh38
rs374311260 Chr.19: 48758114-48758114 on Build GRCh38
rs371287829 Chr.19: 48758019-48758019 on Build GRCh38
rs1009354409 Chr.19: 48758056-48758056 on Build GRCh38
rs756462683 Chr.19: 48758170-48758170 on Build GRCh38
rs754101733 Chr.19: 48758044-48758044 on Build GRCh38
rs772658085 Chr.19: 48758069-48758069 on Build GRCh38
rs1051839058 Chr.19: 48758183-48758183 on Build GRCh38
rs374501551 Chr.19: 48758078-48758078 on Build GRCh38
rs3745708 Chr.19: 48758123-48758123 on Build GRCh38
rs983618958 Chr.19: 48758147-48758147 on Build GRCh38
rs772658968 Chr.19: 48758061-48758061 on Build GRCh38
rs371068639 Chr.19: 48758066-48758066 on Build GRCh38
rs780439354 Chr.19: 48758062-48758062 on Build GRCh38
rs752583057 Chr.19: 48758102-48758102 on Build GRCh38
rs758403141 Chr.19: 48758105-48758105 on Build GRCh38
rs759663403 Chr.19: 48758213-48758213 on Build GRCh38
rs765579156 Chr.19: 48758104-48758104 on Build GRCh38
rs571891800 Chr.19: 48758207-48758207 on Build GRCh38
rs776608911 Chr.19: 48758206-48758206 on Build GRCh38
rs748872169 Chr.19: 48758177-48758177 on Build GRCh38
rs767522194 Chr.19: 48758133-48758133 on Build GRCh38
rs1023366244 Chr.19: 48758084-48758084 on Build GRCh38
rs762834260 Chr.19: 48758018-48758018 on Build GRCh38
rs376996120 Chr.19: 48758107-48758107 on Build GRCh38
rs772168454 Chr.19: 48758101-48758101 on Build GRCh38
rs144978172 Chr.19: 48758014-48758014 on Build GRCh38
rs775575253 Chr.19: 48758194-48758194 on Build GRCh38
rs750659131 Chr.19: 48758143-48758143 on Build GRCh38
rs368200691 Chr.19: 48758115-48758115 on Build GRCh38
rs760751014 Chr.19: 48758197-48758197 on Build GRCh38
rs199499120 Chr.19: 48758034-48758034 on Build GRCh38
rs838130 Chr.19: 48758211-48758211 on Build GRCh38
rs778495177 Chr.19: 48758189-48758189 on Build GRCh38

View Full Product Details