Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 168906449-168906720 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
G6PC2
Entrez Gene ID
57818
Gene Name
glucose-6-phosphatase catalytic subunit 2
Gene Aliases
IGRP
UniGene
Hs.283963
Gene Chromosome Location
Chr.2: 168901223-168910000 on Build GRCh38
Transcripts
NM_001081686.1 NM_021176.2 XM_011511564.2 XM_011511565.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1400995 c.457C>A p.L153I Chr.2: 168906680-168906680 on Build GRCh38
4086538 c.489T>A p.S163R Chr.2: 168906712-168906712 on Build GRCh38
1325774 c.493T>C p.C165R Chr.2: 168906716-168906716 on Build GRCh38
1631432 c.447C>A p.T149T Chr.2: 168906670-168906670 on Build GRCh38
5261112 c.462G>C p.W154C Chr.2: 168906685-168906685 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs997529629 Chr.2: 168906687-168906687 on Build GRCh38
rs539102106 Chr.2: 168906626-168906626 on Build GRCh38
rs773371491 Chr.2: 168906676-168906676 on Build GRCh38
rs756521075 Chr.2: 168906692-168906692 on Build GRCh38
rs780583478 Chr.2: 168906614-168906614 on Build GRCh38
rs765398858 Chr.2: 168906673-168906673 on Build GRCh38
rs534435353 Chr.2: 168906490-168906490 on Build GRCh38
rs970743466 Chr.2: 168906549-168906549 on Build GRCh38
rs536605751 Chr.2: 168906568-168906568 on Build GRCh38
rs914932675 Chr.2: 168906542-168906542 on Build GRCh38
rs775468067 Chr.2: 168906663-168906663 on Build GRCh38
rs201602027 Chr.2: 168906622-168906622 on Build GRCh38
rs189359834 Chr.2: 168906582-168906582 on Build GRCh38
rs766672040 Chr.2: 168906682-168906682 on Build GRCh38
rs2232321 Chr.2: 168906639-168906639 on Build GRCh38
rs560887 Chr.2: 168906638-168906638 on Build GRCh38
rs181737385 Chr.2: 168906660-168906660 on Build GRCh38
rs1026774674 Chr.2: 168906534-168906534 on Build GRCh38
rs150603127 Chr.2: 168906500-168906500 on Build GRCh38
rs761866606 Chr.2: 168906666-168906666 on Build GRCh38
rs751915325 Chr.2: 168906684-168906684 on Build GRCh38
rs745625202 Chr.2: 168906659-168906659 on Build GRCh38
rs774161448 Chr.2: 168906644-168906644 on Build GRCh38
rs780381079 Chr.2: 168906648-168906648 on Build GRCh38
rs899224439 Chr.2: 168906523-168906523 on Build GRCh38
rs937645696 Chr.2: 168906525-168906525 on Build GRCh38
rs755335802 Chr.2: 168906686-168906686 on Build GRCh38
rs753230056 Chr.2: 168906691-168906691 on Build GRCh38
rs770502866 Chr.2: 168906643-168906643 on Build GRCh38
rs567961075 Chr.2: 168906524-168906524 on Build GRCh38
rs767972702 Chr.2: 168906688-168906688 on Build GRCh38
rs368697574 Chr.2: 168906640-168906640 on Build GRCh38

View Full Product Details