Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 10538381-10538642 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRSS55
Entrez Gene ID
203074
Gene Name
protease, serine 55
Gene Aliases
CT153, T-SP1, TSP1, UNQ9391
UniGene
Hs.591394
Gene Chromosome Location
Chr.8: 10525331-10554166 on Build GRCh38
Transcripts
NM_001197020.1 NM_198464.3 XM_005272372.3 XM_011543814.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4406974 c.742-3C>A p.? Chr.8: 10538473-10538473 on Build GRCh38
6515967 c.747C>A p.D249E Chr.8: 10538481-10538481 on Build GRCh38
7385816 c.834G>C p.E278D Chr.8: 10538568-10538568 on Build GRCh38
3643379 c.756G>A p.G252G Chr.8: 10538490-10538490 on Build GRCh38
6039107 c.779C>T p.P260L Chr.8: 10538513-10538513 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764471112 Chr.8: 10538480-10538480 on Build GRCh38
rs200500250 Chr.8: 10538512-10538512 on Build GRCh38
rs546477008 Chr.8: 10538620-10538620 on Build GRCh38
rs754528938 Chr.8: 10538564-10538564 on Build GRCh38
rs776714202 Chr.8: 10538477-10538477 on Build GRCh38
rs778498932 Chr.8: 10538565-10538565 on Build GRCh38
rs73195184 Chr.8: 10538613-10538613 on Build GRCh38
rs147154421 Chr.8: 10538502-10538502 on Build GRCh38
rs372320879 Chr.8: 10538534-10538534 on Build GRCh38
rs751829343 Chr.8: 10538484-10538485 on Build GRCh38
rs776995885 Chr.8: 10538530-10538530 on Build GRCh38
rs780977354 Chr.8: 10538495-10538495 on Build GRCh38
rs974478714 Chr.8: 10538594-10538594 on Build GRCh38
rs1133417 Chr.8: 10538568-10538568 on Build GRCh38
rs953559494 Chr.8: 10538548-10538548 on Build GRCh38
rs768540446 Chr.8: 10538425-10538425 on Build GRCh38
rs141414987 Chr.8: 10538492-10538492 on Build GRCh38
rs761755954 Chr.8: 10538538-10538538 on Build GRCh38
rs746223991 Chr.8: 10538576-10538576 on Build GRCh38
rs755263101 Chr.8: 10538543-10538543 on Build GRCh38
rs753689153 Chr.8: 10538612-10538612 on Build GRCh38
rs748825662 Chr.8: 10538462-10538462 on Build GRCh38
rs370326919 Chr.8: 10538441-10538441 on Build GRCh38
rs752733786 Chr.8: 10538487-10538487 on Build GRCh38
rs771345596 Chr.8: 10538529-10538529 on Build GRCh38
rs61743179 Chr.8: 10538546-10538546 on Build GRCh38
rs201692166 Chr.8: 10538468-10538468 on Build GRCh38
rs749207077 Chr.8: 10538603-10538618 on Build GRCh38
rs759849898 Chr.8: 10538532-10538532 on Build GRCh38
rs368737166 Chr.8: 10538406-10538406 on Build GRCh38
rs750374033 Chr.8: 10538595-10538595 on Build GRCh38
rs781590784 Chr.8: 10538574-10538574 on Build GRCh38
rs765401698 Chr.8: 10538483-10538483 on Build GRCh38
rs777721498 Chr.8: 10538442-10538442 on Build GRCh38
rs375285603 Chr.8: 10538443-10538443 on Build GRCh38
rs754274442 Chr.8: 10538523-10538523 on Build GRCh38
rs970742975 Chr.8: 10538428-10538428 on Build GRCh38
rs748998845 Chr.8: 10538408-10538408 on Build GRCh38
rs962155910 Chr.8: 10538538-10538540 on Build GRCh38
rs758514798 Chr.8: 10538489-10538489 on Build GRCh38
rs751559292 Chr.8: 10538490-10538490 on Build GRCh38
rs774369842 Chr.8: 10538584-10538584 on Build GRCh38
rs374152524 Chr.8: 10538445-10538445 on Build GRCh38
rs56141302 Chr.8: 10538473-10538473 on Build GRCh38
rs950555223 Chr.8: 10538464-10538464 on Build GRCh38
rs768327437 Chr.8: 10538507-10538507 on Build GRCh38
rs140412227 Chr.8: 10538506-10538506 on Build GRCh38
rs377316926 Chr.8: 10538573-10538573 on Build GRCh38
rs760502855 Chr.8: 10538596-10538596 on Build GRCh38
rs142994737 Chr.8: 10538553-10538553 on Build GRCh38
rs763096976 Chr.8: 10538581-10538581 on Build GRCh38
rs757422393 Chr.8: 10538567-10538569 on Build GRCh38
rs780698494 Chr.8: 10538449-10538449 on Build GRCh38
rs190614928 Chr.8: 10538547-10538547 on Build GRCh38
rs769340092 Chr.8: 10538451-10538451 on Build GRCh38
rs745667490 Chr.8: 10538501-10538501 on Build GRCh38
rs770204100 Chr.8: 10538578-10538578 on Build GRCh38
rs6601483 Chr.8: 10538544-10538544 on Build GRCh38
rs141017851 Chr.8: 10538583-10538583 on Build GRCh38
rs765424360 Chr.8: 10538426-10538426 on Build GRCh38
rs753600571 Chr.8: 10538556-10538556 on Build GRCh38
rs764227315 Chr.8: 10538535-10538535 on Build GRCh38
rs745604965 Chr.8: 10538450-10538450 on Build GRCh38
rs752222001 Chr.8: 10538615-10538615 on Build GRCh38
rs538394402 Chr.8: 10538409-10538409 on Build GRCh38
rs550663063 Chr.8: 10538494-10538494 on Build GRCh38
rs775739468 Chr.8: 10538533-10538533 on Build GRCh38
rs773892775 Chr.8: 10538469-10538469 on Build GRCh38
rs766342276 Chr.8: 10538611-10538611 on Build GRCh38
rs894308732 Chr.8: 10538504-10538504 on Build GRCh38
rs775688348 Chr.8: 10538579-10538579 on Build GRCh38
rs761826111 Chr.8: 10538585-10538585 on Build GRCh38
rs144166129 Chr.8: 10538537-10538537 on Build GRCh38
rs375716099 Chr.8: 10538478-10538478 on Build GRCh38
rs781278378 Chr.8: 10538595-10538600 on Build GRCh38
rs1050682091 Chr.8: 10538458-10538458 on Build GRCh38
rs753193702 Chr.8: 10538619-10538619 on Build GRCh38
rs771429634 Chr.8: 10538571-10538571 on Build GRCh38
rs200532088 Chr.8: 10538508-10538508 on Build GRCh38
rs767454552 Chr.8: 10538591-10538591 on Build GRCh38

View Full Product Details