Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33349402-33349655 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFNAR1
Entrez Gene ID
3454
Gene Name
interferon alpha and beta receptor subunit 1
Gene Aliases
AVP, IFN-alpha-REC, IFNAR, IFNBR, IFRC
UniGene
Hs.529400
Gene Chromosome Location
Chr.21: 33324443-33360361 on Build GRCh38
Transcripts
NM_000629.2 XM_005260964.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5642559 c.1048A>G p.I350V Chr.21: 33349448-33349448 on Build GRCh38
1307741 c.1008C>G p.V336V Chr.21: 33349408-33349408 on Build GRCh38
229375 c.1039C>T p.H347Y Chr.21: 33349439-33349439 on Build GRCh38
4935268 c.1041T>C p.H347H Chr.21: 33349441-33349441 on Build GRCh38
6026658 c.1021T>C p.S341P Chr.21: 33349421-33349421 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139423088 Chr.21: 33349451-33349451 on Build GRCh38
rs199515722 Chr.21: 33349511-33349511 on Build GRCh38
rs151316665 Chr.21: 33349450-33349450 on Build GRCh38
rs770527110 Chr.21: 33349555-33349559 on Build GRCh38
rs779819616 Chr.21: 33349591-33349591 on Build GRCh38
rs760075651 Chr.21: 33349531-33349531 on Build GRCh38
rs369150603 Chr.21: 33349440-33349440 on Build GRCh38
rs776376265 Chr.21: 33349510-33349510 on Build GRCh38
rs778751903 Chr.21: 33349567-33349567 on Build GRCh38
rs745587790 Chr.21: 33349574-33349574 on Build GRCh38
rs373432616 Chr.21: 33349477-33349477 on Build GRCh38
rs757107786 Chr.21: 33349565-33349565 on Build GRCh38
rs755054544 Chr.21: 33349432-33349432 on Build GRCh38
rs768458064 Chr.21: 33349509-33349509 on Build GRCh38
rs770688662 Chr.21: 33349460-33349460 on Build GRCh38
rs772055172 Chr.21: 33349584-33349584 on Build GRCh38
rs912613786 Chr.21: 33349600-33349600 on Build GRCh38
rs760199149 Chr.21: 33349506-33349506 on Build GRCh38
rs766737022 Chr.21: 33349529-33349529 on Build GRCh38
rs752074349 Chr.21: 33349530-33349530 on Build GRCh38
rs756176280 Chr.21: 33349581-33349586 on Build GRCh38
rs775083329 Chr.21: 33349491-33349491 on Build GRCh38
rs977884960 Chr.21: 33349483-33349483 on Build GRCh38
rs750514503 Chr.21: 33349574-33349576 on Build GRCh38
rs767035223 Chr.21: 33349480-33349480 on Build GRCh38
rs767845835 Chr.21: 33349532-33349532 on Build GRCh38
rs989832176 Chr.21: 33349516-33349516 on Build GRCh38
rs199669022 Chr.21: 33349555-33349555 on Build GRCh38
rs781328296 Chr.21: 33349435-33349435 on Build GRCh38
rs774191862 Chr.21: 33349463-33349463 on Build GRCh38
rs72549221 Chr.21: 33349582-33349583 on Build GRCh38
rs944140314 Chr.21: 33349613-33349613 on Build GRCh38
rs765920686 Chr.21: 33349556-33349556 on Build GRCh38
rs17875834 Chr.21: 33349476-33349476 on Build GRCh38
rs200354866 Chr.21: 33349539-33349539 on Build GRCh38
rs984843845 Chr.21: 33349489-33349489 on Build GRCh38
rs756137377 Chr.21: 33349544-33349544 on Build GRCh38
rs1020198639 Chr.21: 33349442-33349442 on Build GRCh38

View Full Product Details