Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 196235809-196236036 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNT2
Entrez Gene ID
343450
Gene Name
potassium sodium-activated channel subfamily T member 2
Gene Aliases
KCa4.2, SLICK, SLO2.1
UniGene
Hs.657046
Gene Chromosome Location
Chr.1: 196225779-196608905 on Build GRCh38
Transcripts
NM_001287819.1 NM_001287820.1 NM_198503.3 XM_006711294.2 XM_006711295.2 XM_011509482.2 XM_017001181.1 XM_017001182.1 XM_017001183.1 XM_017001184.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4026448 c.3098+2T>C p.? Chr.1: 196235984-196235984 on Build GRCh38
6123243 c.3249C>A p.I1083I Chr.1: 196236033-196236033 on Build GRCh38
5596910 c.3250C>T p.L1084L Chr.1: 196236032-196236032 on Build GRCh38
5378993 c.3289G>A p.D1097N Chr.1: 196235993-196235993 on Build GRCh38
5759011 c.3280G>C p.E1094Q Chr.1: 196236002-196236002 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs972962900 Chr.1: 196235923-196235923 on Build GRCh38
rs895271619 Chr.1: 196235886-196235886 on Build GRCh38
rs939362677 Chr.1: 196235879-196235879 on Build GRCh38
rs375199969 Chr.1: 196235961-196235961 on Build GRCh38
rs573616600 Chr.1: 196236010-196236010 on Build GRCh38
rs112896769 Chr.1: 196235864-196235864 on Build GRCh38
rs1018755238 Chr.1: 196235856-196235856 on Build GRCh38
rs910129429 Chr.1: 196235914-196235914 on Build GRCh38
rs779810555 Chr.1: 196235962-196235966 on Build GRCh38
rs1056553538 Chr.1: 196235882-196235882 on Build GRCh38
rs539347760 Chr.1: 196235846-196235846 on Build GRCh38
rs558124452 Chr.1: 196235912-196235912 on Build GRCh38
rs201788126 Chr.1: 196235955-196235955 on Build GRCh38
rs368828604 Chr.1: 196235852-196235852 on Build GRCh38
rs770424892 Chr.1: 196235968-196235968 on Build GRCh38
rs948136681 Chr.1: 196235889-196235890 on Build GRCh38
rs771948142 Chr.1: 196235938-196235938 on Build GRCh38
rs747981032 Chr.1: 196235867-196235867 on Build GRCh38
rs984495611 Chr.1: 196235910-196235910 on Build GRCh38
rs371861098 Chr.1: 196235956-196235956 on Build GRCh38
rs776270024 Chr.1: 196235976-196235976 on Build GRCh38
rs111707109 Chr.1: 196235939-196235939 on Build GRCh38
rs887954552 Chr.1: 196235918-196235918 on Build GRCh38
rs921713183 Chr.1: 196235878-196235878 on Build GRCh38
rs951786417 Chr.1: 196235894-196235894 on Build GRCh38
rs928114065 Chr.1: 196235925-196235925 on Build GRCh38
rs759132983 Chr.1: 196235987-196235987 on Build GRCh38
rs940244888 Chr.1: 196235916-196235916 on Build GRCh38

View Full Product Details