Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 85878160-85878421 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NTS
Entrez Gene ID
4922
Gene Name
neurotensin
Gene Aliases
NMN-125, NN, NT, NT/N, NTS1
UniGene
Hs.80962
Gene Chromosome Location
Chr.12: 85874295-85882992 on Build GRCh38
Transcripts
NM_006183.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6562567 c.136-3C>G p.? Chr.12: 85878342-85878342 on Build GRCh38
7084136 c.207_211delCAGCC p.P71fs*1 Chr.12: 85878416-85878420 on Build GRCh38
3793013 c.172C>G p.L58V Chr.12: 85878381-85878381 on Build GRCh38
3793014 c.185G>T p.C62F Chr.12: 85878394-85878394 on Build GRCh38
5904383 c.196A>G p.N66D Chr.12: 85878405-85878405 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762555706 Chr.12: 85878321-85878321 on Build GRCh38
rs190423355 Chr.12: 85878332-85878332 on Build GRCh38
rs768633762 Chr.12: 85878306-85878306 on Build GRCh38
rs764965721 Chr.12: 85878324-85878325 on Build GRCh38
rs779250818 Chr.12: 85878248-85878248 on Build GRCh38
rs779179865 Chr.12: 85878358-85878358 on Build GRCh38
rs762300197 Chr.12: 85878326-85878326 on Build GRCh38
rs533636101 Chr.12: 85878325-85878326 on Build GRCh38
rs563214288 Chr.12: 85878310-85878310 on Build GRCh38
rs924408493 Chr.12: 85878250-85878250 on Build GRCh38
rs753921708 Chr.12: 85878342-85878342 on Build GRCh38
rs988322236 Chr.12: 85878384-85878384 on Build GRCh38
rs185134368 Chr.12: 85878301-85878301 on Build GRCh38
rs977257473 Chr.12: 85878230-85878230 on Build GRCh38
rs746778477 Chr.12: 85878305-85878305 on Build GRCh38
rs780920458 Chr.12: 85878375-85878375 on Build GRCh38
rs759992678 Chr.12: 85878326-85878326 on Build GRCh38
rs960866607 Chr.12: 85878377-85878377 on Build GRCh38
rs1007889489 Chr.12: 85878393-85878393 on Build GRCh38
rs992224201 Chr.12: 85878311-85878311 on Build GRCh38
rs527829573 Chr.12: 85878324-85878324 on Build GRCh38
rs761580952 Chr.12: 85878221-85878221 on Build GRCh38
rs762513562 Chr.12: 85878387-85878387 on Build GRCh38
rs772666850 Chr.12: 85878382-85878382 on Build GRCh38
rs138660407 Chr.12: 85878319-85878319 on Build GRCh38
rs869025243 Chr.12: 85878394-85878394 on Build GRCh38
rs897801727 Chr.12: 85878304-85878304 on Build GRCh38
rs757484576 Chr.12: 85878349-85878349 on Build GRCh38
rs768439469 Chr.12: 85878363-85878363 on Build GRCh38
rs769743232 Chr.12: 85878376-85878376 on Build GRCh38
rs756638931 Chr.12: 85878335-85878335 on Build GRCh38
rs995226412 Chr.12: 85878308-85878308 on Build GRCh38
rs997279600 Chr.12: 85878334-85878334 on Build GRCh38
rs200522935 Chr.12: 85878325-85878325 on Build GRCh38
rs1035109210 Chr.12: 85878369-85878369 on Build GRCh38
rs533444793 Chr.12: 85878318-85878318 on Build GRCh38
rs947014297 Chr.12: 85878391-85878391 on Build GRCh38

View Full Product Details