Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42544898-42545153 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC57
Entrez Gene ID
255252
Gene Name
leucine rich repeat containing 57
Gene Aliases
-
UniGene
Hs.234681
Gene Chromosome Location
Chr.15: 42528332-42548804 on Build GRCh38
Transcripts
NM_153260.2 XM_011521423.2 XM_011521424.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
212031 c.610T>C p.C204R Chr.15: 42545145-42545145 on Build GRCh38
961750 c.655C>T p.R219* Chr.15: 42545100-42545100 on Build GRCh38
1562928 c.635T>A p.L212H Chr.15: 42545120-42545120 on Build GRCh38
6703989 c.620C>T p.A207V Chr.15: 42545135-42545135 on Build GRCh38
700559 c.673G>C p.D225H Chr.15: 42545082-42545082 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760712110 Chr.15: 42545050-42545050 on Build GRCh38
rs764036765 Chr.15: 42545055-42545055 on Build GRCh38
rs1007239371 Chr.15: 42545107-42545107 on Build GRCh38
rs775116351 Chr.15: 42545047-42545047 on Build GRCh38
rs373083935 Chr.15: 42545099-42545099 on Build GRCh38
rs980054192 Chr.15: 42544976-42544976 on Build GRCh38
rs767413509 Chr.15: 42545043-42545043 on Build GRCh38
rs747458901 Chr.15: 42544984-42544984 on Build GRCh38
rs935902669 Chr.15: 42545003-42545003 on Build GRCh38
rs780511238 Chr.15: 42545109-42545109 on Build GRCh38
rs796764778 Chr.15: 42544959-42544959 on Build GRCh38
rs796898892 Chr.15: 42545051-42545052 on Build GRCh38
rs770015336 Chr.15: 42545041-42545041 on Build GRCh38
rs867827960 Chr.15: 42545125-42545125 on Build GRCh38
rs773308767 Chr.15: 42545068-42545068 on Build GRCh38
rs753733118 Chr.15: 42545057-42545057 on Build GRCh38
rs897684120 Chr.15: 42545005-42545005 on Build GRCh38
rs1053452486 Chr.15: 42545037-42545037 on Build GRCh38
rs187496385 Chr.15: 42544999-42544999 on Build GRCh38
rs185145022 Chr.15: 42544944-42544944 on Build GRCh38
rs750150624 Chr.15: 42545100-42545100 on Build GRCh38
rs191625839 Chr.15: 42545115-42545115 on Build GRCh38
rs755394072 Chr.15: 42545046-42545046 on Build GRCh38
rs1056774581 Chr.15: 42545000-42545000 on Build GRCh38
rs552904740 Chr.15: 42545079-42545079 on Build GRCh38
rs369657844 Chr.15: 42545061-42545061 on Build GRCh38
rs948836216 Chr.15: 42544951-42544951 on Build GRCh38
rs867848424 Chr.15: 42545126-42545126 on Build GRCh38

View Full Product Details