Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 56177238-56177463 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR5J2
Entrez Gene ID
282775
Gene Name
olfactory receptor family 5 subfamily J member 2
Gene Aliases
OR11-266
UniGene
Hs.537145
Gene Chromosome Location
Chr.11: 56176618-56177556 on Build GRCh38
Transcripts
NM_001005492.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6537351 c.836C>G p.T279R Chr.11: 56177453-56177453 on Build GRCh38
313730 c.728C>T p.S243F Chr.11: 56177345-56177345 on Build GRCh38
7245692 c.680G>C p.R227T Chr.11: 56177297-56177297 on Build GRCh38
6552061 c.758G>T p.G253V Chr.11: 56177375-56177375 on Build GRCh38
4033822 c.842G>T p.G281V Chr.11: 56177459-56177459 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs536548848 Chr.11: 56177339-56177339 on Build GRCh38
rs757880597 Chr.11: 56177376-56177376 on Build GRCh38
rs757035260 Chr.11: 56177272-56177272 on Build GRCh38
rs142761104 Chr.11: 56177315-56177315 on Build GRCh38
rs760717579 Chr.11: 56177332-56177332 on Build GRCh38
rs779964381 Chr.11: 56177390-56177390 on Build GRCh38
rs745595664 Chr.11: 56177316-56177316 on Build GRCh38
rs755394606 Chr.11: 56177287-56177287 on Build GRCh38
rs961561145 Chr.11: 56177270-56177270 on Build GRCh38
rs760890519 Chr.11: 56177341-56177342 on Build GRCh38
rs140282757 Chr.11: 56177303-56177303 on Build GRCh38
rs753110839 Chr.11: 56177357-56177357 on Build GRCh38
rs756713357 Chr.11: 56177368-56177368 on Build GRCh38
rs753399221 Chr.11: 56177265-56177265 on Build GRCh38
rs771934315 Chr.11: 56177321-56177321 on Build GRCh38
rs750127531 Chr.11: 56177343-56177343 on Build GRCh38
rs766836725 Chr.11: 56177277-56177277 on Build GRCh38
rs139886298 Chr.11: 56177384-56177384 on Build GRCh38
rs903404369 Chr.11: 56177294-56177294 on Build GRCh38
rs755244934 Chr.11: 56177354-56177354 on Build GRCh38
rs868722629 Chr.11: 56177293-56177293 on Build GRCh38
rs199679290 Chr.11: 56177291-56177291 on Build GRCh38
rs757338200 Chr.11: 56177378-56177378 on Build GRCh38
rs376303545 Chr.11: 56177417-56177417 on Build GRCh38
rs768579526 Chr.11: 56177394-56177394 on Build GRCh38
rs868232221 Chr.11: 56177428-56177428 on Build GRCh38
rs201477399 Chr.11: 56177338-56177338 on Build GRCh38
rs150737659 Chr.11: 56177371-56177371 on Build GRCh38
rs554524924 Chr.11: 56177383-56177383 on Build GRCh38
rs756122621 Chr.11: 56177301-56177301 on Build GRCh38
rs781507139 Chr.11: 56177289-56177289 on Build GRCh38
rs771204046 Chr.11: 56177312-56177312 on Build GRCh38
rs767970353 Chr.11: 56177356-56177356 on Build GRCh38
rs751914437 Chr.11: 56177280-56177280 on Build GRCh38
rs777332064 Chr.11: 56177425-56177425 on Build GRCh38
rs147148092 Chr.11: 56177302-56177302 on Build GRCh38
rs866231331 Chr.11: 56177334-56177334 on Build GRCh38
rs568231265 Chr.11: 56177327-56177327 on Build GRCh38
rs776338136 Chr.11: 56177337-56177337 on Build GRCh38
rs763505261 Chr.11: 56177333-56177333 on Build GRCh38
rs915845433 Chr.11: 56177340-56177340 on Build GRCh38
rs149835720 Chr.11: 56177405-56177405 on Build GRCh38
rs140193705 Chr.11: 56177412-56177412 on Build GRCh38
rs1046323612 Chr.11: 56177264-56177264 on Build GRCh38
rs950287067 Chr.11: 56177263-56177263 on Build GRCh38
rs548534792 Chr.11: 56177323-56177323 on Build GRCh38
rs762704004 Chr.11: 56177426-56177426 on Build GRCh38
rs771048735 Chr.11: 56177389-56177389 on Build GRCh38
rs752261661 Chr.11: 56177375-56177375 on Build GRCh38
rs772974358 Chr.11: 56177419-56177419 on Build GRCh38

View Full Product Details