Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 151067316-151067551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MLLT11
Entrez Gene ID
10962
Gene Name
myeloid/lymphoid or mixed-lineage leukemia; translocated to, 11
Gene Aliases
AF1Q
UniGene
Hs.75823
Gene Chromosome Location
Chr.1: 151059675-151068497 on Build GRCh38
Transcripts
NM_006818.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6713375 c.132C>A p.S44R Chr.1: 151067356-151067356 on Build GRCh38
6566686 c.204G>C p.E68D Chr.1: 151067428-151067428 on Build GRCh38
6378044 c.115A>C p.K39Q Chr.1: 151067339-151067339 on Build GRCh38
1257806 c.198C>T p.L66L Chr.1: 151067422-151067422 on Build GRCh38
6310470 c.212C>T p.T71I Chr.1: 151067436-151067436 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752342112 Chr.1: 151067440-151067440 on Build GRCh38
rs753406205 Chr.1: 151067461-151067461 on Build GRCh38
rs773749191 Chr.1: 151067522-151067522 on Build GRCh38
rs778765569 Chr.1: 151067423-151067425 on Build GRCh38
rs587658634 Chr.1: 151067364-151067364 on Build GRCh38
rs763258942 Chr.1: 151067419-151067419 on Build GRCh38
rs111536089 Chr.1: 151067422-151067422 on Build GRCh38
rs11554231 Chr.1: 151067523-151067523 on Build GRCh38
rs587673613 Chr.1: 151067452-151067452 on Build GRCh38
rs753934601 Chr.1: 151067507-151067508 on Build GRCh38
rs776775131 Chr.1: 151067500-151067500 on Build GRCh38
rs587630291 Chr.1: 151067494-151067494 on Build GRCh38
rs148333619 Chr.1: 151067372-151067372 on Build GRCh38
rs11554227 Chr.1: 151067457-151067457 on Build GRCh38
rs11554228 Chr.1: 151067349-151067349 on Build GRCh38
rs776687359 Chr.1: 151067383-151067383 on Build GRCh38
rs76264968 Chr.1: 151067479-151067479 on Build GRCh38
rs752355117 Chr.1: 151067345-151067345 on Build GRCh38
rs142344417 Chr.1: 151067369-151067369 on Build GRCh38
rs755781880 Chr.1: 151067447-151067447 on Build GRCh38
rs779022320 Chr.1: 151067472-151067472 on Build GRCh38
rs201816832 Chr.1: 151067530-151067530 on Build GRCh38
rs773661431 Chr.1: 151067394-151067394 on Build GRCh38
rs1035923911 Chr.1: 151067507-151067507 on Build GRCh38
rs779882393 Chr.1: 151067486-151067486 on Build GRCh38
rs998737563 Chr.1: 151067493-151067493 on Build GRCh38
rs770990780 Chr.1: 151067358-151067358 on Build GRCh38
rs769826203 Chr.1: 151067516-151067516 on Build GRCh38
rs777404639 Chr.1: 151067355-151067355 on Build GRCh38
rs765745252 Chr.1: 151067389-151067389 on Build GRCh38
rs745784490 Chr.1: 151067473-151067473 on Build GRCh38
rs760925045 Chr.1: 151067371-151067371 on Build GRCh38
rs747261433 Chr.1: 151067491-151067491 on Build GRCh38
rs755620628 Chr.1: 151067348-151067348 on Build GRCh38
rs41266626 Chr.1: 151067437-151067437 on Build GRCh38
rs761812866 Chr.1: 151067387-151067387 on Build GRCh38
rs748248042 Chr.1: 151067509-151067509 on Build GRCh38
rs1042817457 Chr.1: 151067467-151067467 on Build GRCh38
rs143015819 Chr.1: 151067470-151067470 on Build GRCh38
rs749300187 Chr.1: 151067357-151067357 on Build GRCh38

View Full Product Details