Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150081175-150081418 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS45
Entrez Gene ID
11311
Gene Name
vacuolar protein sorting 45 homolog
Gene Aliases
H1, H1VPS45, SCN5, VPS45A, VPS45B, VPS54A, VSP45, VSP45A
UniGene
Hs.443750
Gene Chromosome Location
Chr.1: 150067293-150145327 on Build GRCh38
Transcripts
NM_001279353.1 NM_001279354.1 NM_001279355.1 NM_007259.4 NR_103998.1 XM_017000164.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4507546 c.749C>T p.S250F Chr.1: 150081403-150081403 on Build GRCh38
4021926 c.428A>G p.D143G Chr.1: 150081397-150081397 on Build GRCh38
6009452 c.373T>G p.W125G Chr.1: 150081342-150081342 on Build GRCh38
6199348 c.422G>A p.R141Q Chr.1: 150081391-150081391 on Build GRCh38
6199349 c.530G>A p.R177Q Chr.1: 150081391-150081391 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs998534250 Chr.1: 150081220-150081220 on Build GRCh38
rs56203821 Chr.1: 150081222-150081222 on Build GRCh38
rs115802109 Chr.1: 150081307-150081307 on Build GRCh38
rs781938011 Chr.1: 150081390-150081390 on Build GRCh38
rs1011070072 Chr.1: 150081239-150081239 on Build GRCh38
rs782351347 Chr.1: 150081375-150081375 on Build GRCh38
rs782017979 Chr.1: 150081240-150081240 on Build GRCh38
rs587716374 Chr.1: 150081372-150081372 on Build GRCh38
rs782495394 Chr.1: 150081340-150081340 on Build GRCh38
rs587756878 Chr.1: 150081224-150081224 on Build GRCh38
rs782700993 Chr.1: 150081347-150081348 on Build GRCh38
rs782197248 Chr.1: 150081353-150081353 on Build GRCh38
rs782377202 Chr.1: 150081369-150081369 on Build GRCh38
rs36089801 Chr.1: 150081324-150081324 on Build GRCh38
rs782676481 Chr.1: 150081365-150081365 on Build GRCh38
rs782058581 Chr.1: 150081315-150081316 on Build GRCh38
rs782720700 Chr.1: 150081297-150081297 on Build GRCh38
rs3818872 Chr.1: 150081391-150081391 on Build GRCh38
rs930457952 Chr.1: 150081399-150081399 on Build GRCh38
rs782269909 Chr.1: 150081366-150081366 on Build GRCh38
rs587643427 Chr.1: 150081221-150081221 on Build GRCh38
rs587614001 Chr.1: 150081316-150081316 on Build GRCh38
rs953620868 Chr.1: 150081270-150081270 on Build GRCh38
rs782548036 Chr.1: 150081317-150081317 on Build GRCh38
rs36004387 Chr.1: 150081324-150081325 on Build GRCh38
rs782808970 Chr.1: 150081324-150081324 on Build GRCh38
rs781871322 Chr.1: 150081326-150081326 on Build GRCh38
rs782600617 Chr.1: 150081342-150081342 on Build GRCh38

View Full Product Details