Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 139137842-139138067 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC26
Entrez Gene ID
79989
Gene Name
tetratricopeptide repeat domain 26
Gene Aliases
DYF13, IFT56, dyf-13
UniGene
Hs.133020
Gene Chromosome Location
Chr.7: 139133744-139191986 on Build GRCh38
Transcripts
NM_001144920.2 NM_001144923.2 NM_001287512.1 NM_001287513.1 NM_001318333.1 NM_001321740.1 NM_001321741.1 NM_001321742.1 NM_024926.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1622414 c.220A>G p.K74E Chr.7: 139137925-139137925 on Build GRCh38
334762 c.149G>A p.R50H Chr.7: 139137854-139137854 on Build GRCh38
421582 c.157G>A p.G53R Chr.7: 139137862-139137862 on Build GRCh38
4777920 c.158G>A p.G53E Chr.7: 139137863-139137863 on Build GRCh38
4467994 c.151C>T p.H51Y Chr.7: 139137856-139137856 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772408690 Chr.7: 139137970-139137970 on Build GRCh38
rs770258927 Chr.7: 139137930-139137930 on Build GRCh38
rs753673614 Chr.7: 139137888-139137888 on Build GRCh38
rs369346229 Chr.7: 139137932-139137932 on Build GRCh38
rs752609072 Chr.7: 139137914-139137914 on Build GRCh38
rs750975485 Chr.7: 139137917-139137917 on Build GRCh38
rs769240600 Chr.7: 139137956-139137956 on Build GRCh38
rs540127775 Chr.7: 139137999-139137999 on Build GRCh38
rs557445765 Chr.7: 139137984-139137984 on Build GRCh38
rs763172693 Chr.7: 139137864-139137864 on Build GRCh38
rs757944086 Chr.7: 139137915-139137915 on Build GRCh38
rs780743028 Chr.7: 139137947-139137947 on Build GRCh38
rs762121208 Chr.7: 139137877-139137877 on Build GRCh38
rs76685969 Chr.7: 139137989-139137989 on Build GRCh38
rs753173631 Chr.7: 139137991-139137992 on Build GRCh38
rs749841277 Chr.7: 139137948-139137948 on Build GRCh38
rs761735802 Chr.7: 139137967-139137967 on Build GRCh38
rs774507051 Chr.7: 139137957-139137957 on Build GRCh38
rs75136008 Chr.7: 139137902-139137902 on Build GRCh38
rs780702763 Chr.7: 139137926-139137926 on Build GRCh38
rs763870506 Chr.7: 139137990-139137990 on Build GRCh38
rs755553320 Chr.7: 139137956-139137956 on Build GRCh38
rs201276292 Chr.7: 139137871-139137871 on Build GRCh38
rs946290621 Chr.7: 139138009-139138009 on Build GRCh38
rs759249322 Chr.7: 139137900-139137900 on Build GRCh38
rs767581727 Chr.7: 139137878-139137878 on Build GRCh38
rs779417670 Chr.7: 139137987-139137988 on Build GRCh38
rs375989701 Chr.7: 139137916-139137916 on Build GRCh38
rs182585893 Chr.7: 139137923-139137923 on Build GRCh38
rs760928893 Chr.7: 139138012-139138012 on Build GRCh38
rs774251113 Chr.7: 139137876-139137876 on Build GRCh38
rs1010404279 Chr.7: 139137912-139137912 on Build GRCh38
rs773294433 Chr.7: 139137977-139137977 on Build GRCh38
rs996395408 Chr.7: 139137947-139137948 on Build GRCh38
rs764986320 Chr.7: 139137988-139137988 on Build GRCh38
rs746299522 Chr.7: 139137927-139137927 on Build GRCh38
rs1003417702 Chr.7: 139138037-139138037 on Build GRCh38
rs973310229 Chr.7: 139138006-139138006 on Build GRCh38
rs7809077 Chr.7: 139138016-139138016 on Build GRCh38
rs765217642 Chr.7: 139137905-139137905 on Build GRCh38

View Full Product Details