Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 130236929-130237202 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZBTB44
Entrez Gene ID
29068
Gene Name
zinc finger and BTB domain containing 44
Gene Aliases
BTBD15, HSPC063, ZNF851
UniGene
Hs.721470
Gene Chromosome Location
Chr.11: 130226677-130314908 on Build GRCh38
Transcripts
NM_001301098.1 NM_001301099.1 NM_014155.4 XM_005271521.4 XM_005271522.4 XM_006718825.3 XM_006718826.3 XM_011542789.2 XM_017017623.1 XM_017017624.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4430269 c.1330A>T p.M444L Chr.11: 130237031-130237031 on Build GRCh38
7104822 c.1336C>T p.R446C Chr.11: 130237025-130237025 on Build GRCh38
7107034 c.1351+1G>C p.? Chr.11: 130237009-130237009 on Build GRCh38
7104821 c.1336C>T p.R446C Chr.11: 130237025-130237025 on Build GRCh38
6825347 c.1343delA p.K448fs*27 Chr.11: 130237018-130237018 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs912191614 Chr.11: 130237173-130237173 on Build GRCh38
rs772041057 Chr.11: 130237113-130237113 on Build GRCh38
rs576936297 Chr.11: 130237153-130237153 on Build GRCh38
rs756494582 Chr.11: 130237024-130237024 on Build GRCh38
rs766474319 Chr.11: 130236972-130236972 on Build GRCh38
rs756753387 Chr.11: 130237107-130237121 on Build GRCh38
rs745408413 Chr.11: 130237111-130237111 on Build GRCh38
rs775346189 Chr.11: 130237125-130237125 on Build GRCh38
rs752016149 Chr.11: 130236974-130236974 on Build GRCh38
rs148074972 Chr.11: 130237158-130237158 on Build GRCh38
rs767954193 Chr.11: 130236995-130236995 on Build GRCh38
rs769534882 Chr.11: 130236968-130236968 on Build GRCh38
rs756789770 Chr.11: 130237096-130237096 on Build GRCh38
rs373305671 Chr.11: 130236994-130236994 on Build GRCh38
rs762636714 Chr.11: 130237177-130237177 on Build GRCh38
rs527818911 Chr.11: 130237178-130237178 on Build GRCh38
rs753105168 Chr.11: 130237000-130237000 on Build GRCh38
rs778396441 Chr.11: 130237121-130237121 on Build GRCh38
rs576431832 Chr.11: 130237176-130237176 on Build GRCh38
rs375724881 Chr.11: 130237136-130237136 on Build GRCh38
rs748954468 Chr.11: 130237070-130237070 on Build GRCh38
rs553965419 Chr.11: 130237105-130237105 on Build GRCh38
rs765480242 Chr.11: 130236950-130236950 on Build GRCh38
rs956578677 Chr.11: 130237115-130237115 on Build GRCh38
rs375863333 Chr.11: 130236990-130236990 on Build GRCh38
rs933319017 Chr.11: 130237104-130237104 on Build GRCh38
rs773432222 Chr.11: 130236954-130236954 on Build GRCh38
rs777510437 Chr.11: 130237035-130237035 on Build GRCh38

View Full Product Details