Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 10457373-10457616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATF7IP2
Entrez Gene ID
80063
Gene Name
activating transcription factor 7 interacting protein 2
Gene Aliases
MCAF2
UniGene
Hs.513343
Gene Chromosome Location
Chr.16: 10386055-10483640 on Build GRCh38
Transcripts
NM_001256160.1 NM_024997.3 NR_045815.1 NR_045816.1 XM_006720953.3 XM_011522666.2 XM_017023705.1 XM_017023706.1 XM_017023707.1 XM_017023708.1 XM_017023709.1 XM_017023710.1 XM_017023711.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4058129 c.1338G>T p.K446N Chr.16: 10457515-10457515 on Build GRCh38
4058128 c.1211C>T p.A404V Chr.16: 10457388-10457388 on Build GRCh38
5967033 c.1237T>A p.S413T Chr.16: 10457414-10457414 on Build GRCh38
3505701 c.1229A>G p.N410S Chr.16: 10457406-10457406 on Build GRCh38
5900317 c.1349A>C p.E450A Chr.16: 10457526-10457526 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775439017 Chr.16: 10457555-10457555 on Build GRCh38
rs760154727 Chr.16: 10457556-10457556 on Build GRCh38
rs772187949 Chr.16: 10457401-10457401 on Build GRCh38
rs768929782 Chr.16: 10457421-10457421 on Build GRCh38
rs375303503 Chr.16: 10457568-10457568 on Build GRCh38
rs748363076 Chr.16: 10457511-10457511 on Build GRCh38
rs763632025 Chr.16: 10457570-10457570 on Build GRCh38
rs780334449 Chr.16: 10457404-10457404 on Build GRCh38
rs746401028 Chr.16: 10457399-10457399 on Build GRCh38
rs529549876 Chr.16: 10457481-10457481 on Build GRCh38
rs143609619 Chr.16: 10457491-10457491 on Build GRCh38
rs200683589 Chr.16: 10457521-10457521 on Build GRCh38
rs763685143 Chr.16: 10457466-10457466 on Build GRCh38
rs759459380 Chr.16: 10457444-10457444 on Build GRCh38
rs761531766 Chr.16: 10457543-10457544 on Build GRCh38
rs781546339 Chr.16: 10457507-10457507 on Build GRCh38
rs751747949 Chr.16: 10457581-10457581 on Build GRCh38
rs371971593 Chr.16: 10457564-10457564 on Build GRCh38
rs148070233 Chr.16: 10457499-10457499 on Build GRCh38
rs747451962 Chr.16: 10457538-10457538 on Build GRCh38
rs141776960 Chr.16: 10457500-10457500 on Build GRCh38
rs1023053450 Chr.16: 10457565-10457565 on Build GRCh38
rs148868599 Chr.16: 10457429-10457429 on Build GRCh38
rs765062218 Chr.16: 10457580-10457581 on Build GRCh38
rs770683200 Chr.16: 10457430-10457430 on Build GRCh38
rs779211793 Chr.16: 10457474-10457474 on Build GRCh38
rs138588090 Chr.16: 10457426-10457426 on Build GRCh38
rs1014385024 Chr.16: 10457540-10457540 on Build GRCh38
rs752146936 Chr.16: 10457465-10457465 on Build GRCh38
rs747094546 Chr.16: 10457414-10457414 on Build GRCh38
rs144631035 Chr.16: 10457515-10457515 on Build GRCh38
rs771931424 Chr.16: 10457532-10457532 on Build GRCh38
rs755134684 Chr.16: 10457504-10457504 on Build GRCh38
rs753480948 Chr.16: 10457470-10457470 on Build GRCh38
rs867532363 Chr.16: 10457454-10457454 on Build GRCh38
rs267604402 Chr.16: 10457471-10457471 on Build GRCh38
rs1020424191 Chr.16: 10457463-10457463 on Build GRCh38
rs370114952 Chr.16: 10457519-10457519 on Build GRCh38
rs776442274 Chr.16: 10457537-10457537 on Build GRCh38
rs764952189 Chr.16: 10457578-10457578 on Build GRCh38
rs774317372 Chr.16: 10457441-10457441 on Build GRCh38
rs149248106 Chr.16: 10457574-10457574 on Build GRCh38
rs200149128 Chr.16: 10457577-10457577 on Build GRCh38
rs763946306 Chr.16: 10457483-10457483 on Build GRCh38
rs767001897 Chr.16: 10457447-10457447 on Build GRCh38
rs770531609 Chr.16: 10457503-10457503 on Build GRCh38

View Full Product Details