Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 122911482-122911755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZBTB6
Entrez Gene ID
10773
Gene Name
zinc finger and BTB domain containing 6
Gene Aliases
ZID, ZNF482
UniGene
Hs.654596
Gene Chromosome Location
Chr.9: 122908056-122913328 on Build GRCh38
Transcripts
NM_006626.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6825452 c.419C>G p.T140S Chr.9: 122911654-122911654 on Build GRCh38
3382416 c.512C>A p.P171H Chr.9: 122911561-122911561 on Build GRCh38
6825454 c.428G>A p.C143Y Chr.9: 122911645-122911645 on Build GRCh38
4742207 c.526T>A p.F176I Chr.9: 122911547-122911547 on Build GRCh38
6356488 c.547A>C p.S183R Chr.9: 122911526-122911526 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755167945 Chr.9: 122911714-122911714 on Build GRCh38
rs970576802 Chr.9: 122911572-122911572 on Build GRCh38
rs758805977 Chr.9: 122911698-122911698 on Build GRCh38
rs752185575 Chr.9: 122911615-122911615 on Build GRCh38
rs368544617 Chr.9: 122911568-122911568 on Build GRCh38
rs897266111 Chr.9: 122911510-122911510 on Build GRCh38
rs780307294 Chr.9: 122911522-122911522 on Build GRCh38
rs755986826 Chr.9: 122911593-122911593 on Build GRCh38
rs780464383 Chr.9: 122911700-122911700 on Build GRCh38
rs762786793 Chr.9: 122911563-122911563 on Build GRCh38
rs145240136 Chr.9: 122911601-122911601 on Build GRCh38
rs769783824 Chr.9: 122911553-122911553 on Build GRCh38
rs370904446 Chr.9: 122911550-122911550 on Build GRCh38
rs772307663 Chr.9: 122911521-122911521 on Build GRCh38
rs375226568 Chr.9: 122911566-122911566 on Build GRCh38
rs765684059 Chr.9: 122911664-122911664 on Build GRCh38
rs190479955 Chr.9: 122911507-122911507 on Build GRCh38
rs374124505 Chr.9: 122911520-122911520 on Build GRCh38
rs747251074 Chr.9: 122911703-122911703 on Build GRCh38
rs779254297 Chr.9: 122911508-122911508 on Build GRCh38
rs746416374 Chr.9: 122911536-122911536 on Build GRCh38
rs776756327 Chr.9: 122911541-122911541 on Build GRCh38
rs773168525 Chr.9: 122911557-122911557 on Build GRCh38
rs750911126 Chr.9: 122911682-122911682 on Build GRCh38
rs893536700 Chr.9: 122911661-122911661 on Build GRCh38
rs377299938 Chr.9: 122911524-122911524 on Build GRCh38
rs766254165 Chr.9: 122911564-122911564 on Build GRCh38
rs757792817 Chr.9: 122911629-122911629 on Build GRCh38
rs768688033 Chr.9: 122911535-122911535 on Build GRCh38

View Full Product Details