Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 159692654-159692885 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SOD2
Entrez Gene ID
6648
Gene Name
superoxide dismutase 2, mitochondrial
Gene Aliases
IPO-B, IPOB, MNSOD, MVCD6, Mn-SOD
UniGene
Hs.487046
Gene Chromosome Location
Chr.6: 159679064-159762529 on Build GRCh38
Transcripts
NM_000636.3 NM_001024465.2 NM_001024466.2 NM_001322814.1 NM_001322815.1 NM_001322816.1 NM_001322817.1 NM_001322819.1 NM_001322820.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5610668 c.45G>A p.P15P Chr.6: 159692842-159692842 on Build GRCh38
5615634 c.148C>A p.H50N Chr.6: 159692739-159692739 on Build GRCh38
7391327 c.67A>G p.R23G Chr.6: 159692820-159692820 on Build GRCh38
4748057 c.199_201delGAG p.E67delE Chr.6: 159692686-159692688 on Build GRCh38
483631 c.171C>A p.A57A Chr.6: 159692716-159692716 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs4880 Chr.6: 159692840-159692840 on Build GRCh38
rs780273236 Chr.6: 159692707-159692707 on Build GRCh38
rs773283190 Chr.6: 159692686-159692688 on Build GRCh38
rs1804451 Chr.6: 159692755-159692755 on Build GRCh38
rs749865952 Chr.6: 159692710-159692710 on Build GRCh38
rs5746096 Chr.6: 159692858-159692858 on Build GRCh38
rs774208220 Chr.6: 159692841-159692841 on Build GRCh38
rs908831517 Chr.6: 159692781-159692781 on Build GRCh38
rs755480422 Chr.6: 159692714-159692714 on Build GRCh38
rs373540824 Chr.6: 159692688-159692688 on Build GRCh38
rs1009373193 Chr.6: 159692782-159692782 on Build GRCh38
rs926931277 Chr.6: 159692847-159692847 on Build GRCh38
rs756387474 Chr.6: 159692793-159692793 on Build GRCh38
rs77799497 Chr.6: 159692865-159692865 on Build GRCh38
rs139336939 Chr.6: 159692836-159692836 on Build GRCh38
rs772998028 Chr.6: 159692861-159692861 on Build GRCh38
rs766576089 Chr.6: 159692797-159692797 on Build GRCh38
rs758729654 Chr.6: 159692820-159692820 on Build GRCh38
rs5746097 Chr.6: 159692690-159692690 on Build GRCh38
rs763954620 Chr.6: 159692756-159692756 on Build GRCh38
rs761782632 Chr.6: 159692855-159692855 on Build GRCh38
rs774968268 Chr.6: 159692742-159692742 on Build GRCh38
rs373689189 Chr.6: 159692828-159692828 on Build GRCh38
rs750654591 Chr.6: 159692792-159692792 on Build GRCh38
rs953038635 Chr.6: 159692720-159692720 on Build GRCh38
rs1043271668 Chr.6: 159692724-159692724 on Build GRCh38
rs1050822396 Chr.6: 159692772-159692772 on Build GRCh38
rs745565425 Chr.6: 159692827-159692827 on Build GRCh38
rs375884951 Chr.6: 159692774-159692774 on Build GRCh38
rs11551080 Chr.6: 159692692-159692692 on Build GRCh38
rs762291409 Chr.6: 159692815-159692816 on Build GRCh38
rs767598315 Chr.6: 159692691-159692691 on Build GRCh38
rs774235683 Chr.6: 159692758-159692758 on Build GRCh38
rs779238114 Chr.6: 159692811-159692811 on Build GRCh38
rs200407278 Chr.6: 159692806-159692806 on Build GRCh38
rs185564053 Chr.6: 159692835-159692835 on Build GRCh38
rs375177938 Chr.6: 159692736-159692736 on Build GRCh38
rs779421501 Chr.6: 159692716-159692716 on Build GRCh38
rs776194516 Chr.6: 159692731-159692731 on Build GRCh38
rs921294182 Chr.6: 159692701-159692701 on Build GRCh38
rs753223739 Chr.6: 159692819-159692819 on Build GRCh38
rs778195973 Chr.6: 159692824-159692824 on Build GRCh38
rs996125245 Chr.6: 159692750-159692750 on Build GRCh38
rs769361629 Chr.6: 159692737-159692737 on Build GRCh38
rs761468407 Chr.6: 159692770-159692770 on Build GRCh38
rs750541818 Chr.6: 159692697-159692697 on Build GRCh38
rs377317109 Chr.6: 159692698-159692698 on Build GRCh38
rs370671213 Chr.6: 159692722-159692722 on Build GRCh38
rs767325836 Chr.6: 159692776-159692776 on Build GRCh38
rs762893766 Chr.6: 159692747-159692747 on Build GRCh38
rs372258485 Chr.6: 159692843-159692843 on Build GRCh38

View Full Product Details