Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30709348-30709621 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SRCAP
Entrez Gene ID
10847
Gene Name
Snf2-related CREBBP activator protein
Gene Aliases
DOMO1, EAF1, FLHS, SWR1
UniGene
Hs.620916
Gene Chromosome Location
Chr.16: 30699141-30740129 on Build GRCh38
Transcripts
NM_006662.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4555907 c.672G>A p.K224K Chr.16: 30709551-30709551 on Build GRCh38
6907655 c.707T>C p.I236T Chr.16: 30709586-30709586 on Build GRCh38
3112815 c.732G>A p.S244S Chr.16: 30709611-30709611 on Build GRCh38
7429395 c.656C>T p.S219F Chr.16: 30709535-30709535 on Build GRCh38
1301877 c.739C>T p.L247L Chr.16: 30709618-30709618 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs775815386 Chr.16: 30709553-30709553 on Build GRCh38
rs201126167 Chr.16: 30709514-30709514 on Build GRCh38
rs539898059 Chr.16: 30709451-30709451 on Build GRCh38
rs779191843 Chr.16: 30709523-30709523 on Build GRCh38
rs751129266 Chr.16: 30709517-30709517 on Build GRCh38
rs754481074 Chr.16: 30709379-30709379 on Build GRCh38
rs570021658 Chr.16: 30709539-30709539 on Build GRCh38
rs558469214 Chr.16: 30709374-30709374 on Build GRCh38
rs964606202 Chr.16: 30709397-30709397 on Build GRCh38
rs937653172 Chr.16: 30709417-30709417 on Build GRCh38
rs745959283 Chr.16: 30709524-30709524 on Build GRCh38
rs914003060 Chr.16: 30709488-30709488 on Build GRCh38
rs753838120 Chr.16: 30709467-30709467 on Build GRCh38
rs1054837442 Chr.16: 30709434-30709434 on Build GRCh38
rs867536860 Chr.16: 30709515-30709515 on Build GRCh38
rs537390523 Chr.16: 30709563-30709563 on Build GRCh38
rs756520870 Chr.16: 30709519-30709519 on Build GRCh38
rs749840397 Chr.16: 30709521-30709521 on Build GRCh38
rs757834088 Chr.16: 30709522-30709522 on Build GRCh38
rs572844193 Chr.16: 30709588-30709588 on Build GRCh38
rs764770396 Chr.16: 30709478-30709478 on Build GRCh38
rs777976051 Chr.16: 30709520-30709520 on Build GRCh38
rs183202677 Chr.16: 30709490-30709490 on Build GRCh38
rs761718840 Chr.16: 30709469-30709469 on Build GRCh38
rs566011412 Chr.16: 30709448-30709448 on Build GRCh38
rs564146349 Chr.16: 30709494-30709494 on Build GRCh38
rs747153085 Chr.16: 30709554-30709554 on Build GRCh38
rs988205885 Chr.16: 30709456-30709456 on Build GRCh38
rs768548516 Chr.16: 30709560-30709560 on Build GRCh38
rs867014564 Chr.16: 30709480-30709480 on Build GRCh38
rs975671728 Chr.16: 30709443-30709443 on Build GRCh38
rs927726783 Chr.16: 30709383-30709383 on Build GRCh38
rs772213094 Chr.16: 30709548-30709548 on Build GRCh38
rs926651280 Chr.16: 30709536-30709536 on Build GRCh38
rs899375654 Chr.16: 30709450-30709450 on Build GRCh38
rs1046790450 Chr.16: 30709492-30709492 on Build GRCh38
rs376499525 Chr.16: 30709493-30709493 on Build GRCh38
rs995095494 Chr.16: 30709474-30709474 on Build GRCh38

View Full Product Details