Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10514797-10515050 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
S1PR5
Entrez Gene ID
53637
Gene Name
sphingosine-1-phosphate receptor 5
Gene Aliases
EDG8, Edg-8, S1P5, SPPR-1, SPPR-2
UniGene
Hs.501561
Gene Chromosome Location
Chr.19: 10512742-10517992 on Build GRCh38
Transcripts
NM_001166215.1 NM_030760.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7108415 c.5A>C p.E2A Chr.19: 10515007-10515007 on Build GRCh38
6149738 c.150C>T p.I50I Chr.19: 10514862-10514862 on Build GRCh38
6038124 c.78C>T p.R26R Chr.19: 10514934-10514934 on Build GRCh38
372919 c.207T>G p.A69A Chr.19: 10514805-10514805 on Build GRCh38
4820678 c.159G>A p.E53E Chr.19: 10514853-10514853 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768031173 Chr.19: 10514891-10514891 on Build GRCh38
rs534220387 Chr.19: 10515016-10515017 on Build GRCh38
rs375435151 Chr.19: 10514990-10514990 on Build GRCh38
rs1024142586 Chr.19: 10514942-10514942 on Build GRCh38
rs955428256 Chr.19: 10514976-10514976 on Build GRCh38
rs147145897 Chr.19: 10514943-10514943 on Build GRCh38
rs777500457 Chr.19: 10514937-10514937 on Build GRCh38
rs369911931 Chr.19: 10514981-10514981 on Build GRCh38
rs373176714 Chr.19: 10514928-10514928 on Build GRCh38
rs768610698 Chr.19: 10514900-10514900 on Build GRCh38
rs377292940 Chr.19: 10514945-10514945 on Build GRCh38
rs143370233 Chr.19: 10514910-10514910 on Build GRCh38
rs769251884 Chr.19: 10514960-10514960 on Build GRCh38
rs911016774 Chr.19: 10514980-10514980 on Build GRCh38
rs751487851 Chr.19: 10514925-10514925 on Build GRCh38
rs1012723054 Chr.19: 10514892-10514892 on Build GRCh38
rs1005534931 Chr.19: 10514864-10514864 on Build GRCh38
rs375015034 Chr.19: 10514895-10514895 on Build GRCh38
rs759371516 Chr.19: 10515011-10515011 on Build GRCh38
rs753991379 Chr.19: 10514824-10514824 on Build GRCh38
rs769870996 Chr.19: 10514982-10514982 on Build GRCh38
rs766177652 Chr.19: 10514918-10514918 on Build GRCh38
rs767440311 Chr.19: 10515012-10515012 on Build GRCh38
rs769527790 Chr.19: 10514888-10514888 on Build GRCh38
rs560472766 Chr.19: 10514944-10514944 on Build GRCh38
rs773755164 Chr.19: 10514983-10514983 on Build GRCh38
rs779930744 Chr.19: 10514850-10514850 on Build GRCh38
rs747753514 Chr.19: 10514884-10514884 on Build GRCh38
rs748215041 Chr.19: 10514977-10514977 on Build GRCh38
rs765446809 Chr.19: 10514914-10514914 on Build GRCh38
rs780815612 Chr.19: 10514881-10514881 on Build GRCh38
rs752608086 Chr.19: 10515016-10515016 on Build GRCh38
rs754391437 Chr.19: 10514877-10514877 on Build GRCh38
rs764246553 Chr.19: 10514815-10514815 on Build GRCh38
rs369192797 Chr.19: 10514913-10514913 on Build GRCh38
rs138019257 Chr.19: 10514908-10514908 on Build GRCh38
rs762160071 Chr.19: 10514833-10514833 on Build GRCh38
rs777023463 Chr.19: 10514965-10514965 on Build GRCh38
rs758207621 Chr.19: 10514849-10514849 on Build GRCh38
rs774173559 Chr.19: 10514988-10514988 on Build GRCh38
rs751506292 Chr.19: 10514858-10514858 on Build GRCh38
rs764906828 Chr.19: 10514835-10514835 on Build GRCh38
rs200580371 Chr.19: 10514870-10514870 on Build GRCh38
rs766907326 Chr.19: 10514933-10514933 on Build GRCh38
rs779400097 Chr.19: 10514889-10514889 on Build GRCh38
rs766160834 Chr.19: 10514985-10514985 on Build GRCh38
rs755602746 Chr.19: 10514936-10514936 on Build GRCh38
rs747607768 Chr.19: 10514897-10514897 on Build GRCh38
rs986657803 Chr.19: 10514979-10514979 on Build GRCh38
rs533204482 Chr.19: 10514916-10514916 on Build GRCh38
rs562669008 Chr.19: 10514843-10514843 on Build GRCh38

View Full Product Details