Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 178713290-178713533 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF354A
Entrez Gene ID
6940
Gene Name
zinc finger protein 354A
Gene Aliases
EZNF, HEL104, HKL1, KID-1, KID1, TCF17
UniGene
Hs.484324
Gene Chromosome Location
Chr.5: 178711521-178730702 on Build GRCh38
Transcripts
NM_001324339.1 NM_005649.2 XM_011534645.2 XM_017009791.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6836670 c.447C>G p.H149Q Chr.5: 178713431-178713431 on Build GRCh38
1233927 c.362A>C p.K121T Chr.5: 178713516-178713516 on Build GRCh38
5498929 c.453_454insA p.I152fs*9 Chr.5: 178713424-178713425 on Build GRCh38
1581960 c.544A>G p.R182G Chr.5: 178713334-178713334 on Build GRCh38
3919788 c.437C>T p.S146L Chr.5: 178713441-178713441 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746801335 Chr.5: 178713418-178713418 on Build GRCh38
rs909865457 Chr.5: 178713375-178713375 on Build GRCh38
rs764918458 Chr.5: 178713383-178713383 on Build GRCh38
rs754398417 Chr.5: 178713475-178713475 on Build GRCh38
rs749493415 Chr.5: 178713344-178713344 on Build GRCh38
rs764525393 Chr.5: 178713471-178713471 on Build GRCh38
rs186479392 Chr.5: 178713341-178713341 on Build GRCh38
rs765002157 Chr.5: 178713330-178713330 on Build GRCh38
rs201921352 Chr.5: 178713345-178713345 on Build GRCh38
rs141789608 Chr.5: 178713435-178713435 on Build GRCh38
rs756665055 Chr.5: 178713403-178713403 on Build GRCh38
rs756148987 Chr.5: 178713340-178713340 on Build GRCh38
rs780206105 Chr.5: 178713470-178713472 on Build GRCh38
rs753305594 Chr.5: 178713337-178713337 on Build GRCh38
rs77501345 Chr.5: 178713419-178713419 on Build GRCh38
rs545687958 Chr.5: 178713407-178713407 on Build GRCh38
rs377403210 Chr.5: 178713350-178713350 on Build GRCh38
rs77426342 Chr.5: 178713412-178713412 on Build GRCh38
rs761605628 Chr.5: 178713319-178713319 on Build GRCh38
rs906419362 Chr.5: 178713470-178713470 on Build GRCh38
rs779947894 Chr.5: 178713417-178713417 on Build GRCh38
rs761700035 Chr.5: 178713408-178713408 on Build GRCh38
rs753116981 Chr.5: 178713397-178713397 on Build GRCh38
rs142035452 Chr.5: 178713334-178713334 on Build GRCh38
rs767996621 Chr.5: 178713388-178713388 on Build GRCh38
rs201770190 Chr.5: 178713499-178713499 on Build GRCh38
rs757320036 Chr.5: 178713481-178713481 on Build GRCh38
rs767755392 Chr.5: 178713438-178713438 on Build GRCh38
rs550120180 Chr.5: 178713376-178713376 on Build GRCh38
rs776230220 Chr.5: 178713367-178713367 on Build GRCh38
rs767410629 Chr.5: 178713411-178713423 on Build GRCh38
rs751832707 Chr.5: 178713331-178713331 on Build GRCh38
rs553580516 Chr.5: 178713384-178713384 on Build GRCh38
rs759806481 Chr.5: 178713386-178713386 on Build GRCh38
rs758096362 Chr.5: 178713416-178713416 on Build GRCh38
rs199874817 Chr.5: 178713352-178713352 on Build GRCh38
rs775878828 Chr.5: 178713445-178713445 on Build GRCh38
rs746164650 Chr.5: 178713415-178713415 on Build GRCh38
rs765394390 Chr.5: 178713503-178713503 on Build GRCh38
rs781494916 Chr.5: 178713335-178713335 on Build GRCh38
rs201784705 Chr.5: 178713357-178713357 on Build GRCh38
rs201214103 Chr.5: 178713408-178713408 on Build GRCh38
rs765352230 Chr.5: 178713368-178713368 on Build GRCh38
rs776716872 Chr.5: 178713421-178713421 on Build GRCh38
rs1037109651 Chr.5: 178713410-178713410 on Build GRCh38
rs778986209 Chr.5: 178713414-178713414 on Build GRCh38
rs756203863 Chr.5: 178713423-178713424 on Build GRCh38
rs778276728 Chr.5: 178713405-178713405 on Build GRCh38
rs79565699 Chr.5: 178713318-178713318 on Build GRCh38
rs747605360 Chr.5: 178713423-178713423 on Build GRCh38
rs775280733 Chr.5: 178713354-178713354 on Build GRCh38
rs761062256 Chr.5: 178713469-178713469 on Build GRCh38
rs370083523 Chr.5: 178713504-178713504 on Build GRCh38
rs769487776 Chr.5: 178713424-178713424 on Build GRCh38
rs146941883 Chr.5: 178713358-178713358 on Build GRCh38
rs147082488 Chr.5: 178713412-178713423 on Build GRCh38
rs772736179 Chr.5: 178713425-178713425 on Build GRCh38
rs751520495 Chr.5: 178713408-178713423 on Build GRCh38

View Full Product Details