Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 33989144-33989383 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMACR
Entrez Gene ID
23600
Gene Name
alpha-methylacyl-CoA racemase
Gene Aliases
AMACRD, CBAS4, RACE, RM
UniGene
Hs.508343
Gene Chromosome Location
Chr.5: 33986986-34008115 on Build GRCh38
Transcripts
NM_001167595.1 NM_014324.5 NM_203382.2 NR_037951.1

Gene Symbol
C1QTNF3-AMACR
Entrez Gene ID
100534612
Gene Name
C1QTNF3-AMACR readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.171929
Gene Chromosome Location
Chr.5: 33986986-34124528 on Build GRCh38
Transcripts
NM_001167595.1 NM_014324.5 NM_203382.2 NR_037951.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1067410 c.1084G>A p.E362K Chr.5: 33989158-33989158 on Build GRCh38
1067411 c.1042G>T p.E348* Chr.5: 33989200-33989200 on Build GRCh38
3732073 c.1007C>A p.A336D Chr.5: 33989235-33989235 on Build GRCh38
1067412 c.887C>T p.P296L Chr.5: 33989355-33989355 on Build GRCh38
5356313 c.976delC p.R326fs*7 Chr.5: 33989266-33989266 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746094231 Chr.5: 33989236-33989236 on Build GRCh38
rs368693738 Chr.5: 33989241-33989241 on Build GRCh38
rs754326212 Chr.5: 33989285-33989285 on Build GRCh38
rs749703699 Chr.5: 33989217-33989217 on Build GRCh38
rs1024198680 Chr.5: 33989235-33989235 on Build GRCh38
rs772946350 Chr.5: 33989329-33989329 on Build GRCh38
rs142574369 Chr.5: 33989242-33989242 on Build GRCh38
rs371343345 Chr.5: 33989266-33989266 on Build GRCh38
rs968981597 Chr.5: 33989336-33989336 on Build GRCh38
rs779290495 Chr.5: 33989231-33989231 on Build GRCh38
rs764961991 Chr.5: 33989264-33989264 on Build GRCh38
rs139973015 Chr.5: 33989300-33989300 on Build GRCh38
rs769705590 Chr.5: 33989324-33989324 on Build GRCh38
rs778402751 Chr.5: 33989211-33989211 on Build GRCh38
rs761467654 Chr.5: 33989277-33989309 on Build GRCh38
rs751045891 Chr.5: 33989202-33989202 on Build GRCh38
rs764977396 Chr.5: 33989193-33989193 on Build GRCh38
rs772219669 Chr.5: 33989239-33989239 on Build GRCh38
rs756743141 Chr.5: 33989205-33989205 on Build GRCh38
rs764967392 Chr.5: 33989313-33989315 on Build GRCh38
rs575263204 Chr.5: 33989277-33989277 on Build GRCh38
rs757726794 Chr.5: 33989290-33989290 on Build GRCh38
rs886060530 Chr.5: 33989353-33989353 on Build GRCh38
rs750174939 Chr.5: 33989265-33989265 on Build GRCh38
rs974623718 Chr.5: 33989354-33989354 on Build GRCh38
rs779154363 Chr.5: 33989291-33989291 on Build GRCh38
rs776398193 Chr.5: 33989257-33989257 on Build GRCh38
rs764011770 Chr.5: 33989173-33989173 on Build GRCh38
rs753686325 Chr.5: 33989176-33989176 on Build GRCh38
rs113361629 Chr.5: 33989298-33989298 on Build GRCh38
rs201507169 Chr.5: 33989308-33989308 on Build GRCh38
rs761768331 Chr.5: 33989258-33989258 on Build GRCh38
rs143542702 Chr.5: 33989363-33989363 on Build GRCh38
rs751164365 Chr.5: 33989270-33989270 on Build GRCh38
rs886060529 Chr.5: 33989305-33989305 on Build GRCh38
rs750073270 Chr.5: 33989194-33989194 on Build GRCh38
rs768752342 Chr.5: 33989303-33989303 on Build GRCh38
rs766116509 Chr.5: 33989267-33989267 on Build GRCh38
rs138052540 Chr.5: 33989172-33989172 on Build GRCh38
rs760727161 Chr.5: 33989249-33989249 on Build GRCh38
rs772334299 Chr.5: 33989299-33989299 on Build GRCh38
rs200505839 Chr.5: 33989275-33989275 on Build GRCh38
rs774180397 Chr.5: 33989355-33989355 on Build GRCh38
rs761488582 Chr.5: 33989191-33989191 on Build GRCh38
rs906797872 Chr.5: 33989276-33989276 on Build GRCh38
rs1055202 Chr.5: 33989262-33989262 on Build GRCh38
rs776723461 Chr.5: 33989307-33989307 on Build GRCh38
rs771232800 Chr.5: 33989229-33989229 on Build GRCh38
rs747129541 Chr.5: 33989301-33989301 on Build GRCh38
rs779722935 Chr.5: 33989197-33989197 on Build GRCh38
rs768627644 Chr.5: 33989253-33989253 on Build GRCh38
rs762820381 Chr.5: 33989332-33989332 on Build GRCh38

View Full Product Details