Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232457580-232457819 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALPI
Entrez Gene ID
248
Gene Name
alkaline phosphatase, intestinal
Gene Aliases
IAP
UniGene
Hs.37009
Gene Chromosome Location
Chr.2: 232456112-232460745 on Build GRCh38
Transcripts
NM_001631.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4661221 c.800G>C p.W267S Chr.2: 232457811-232457811 on Build GRCh38
3049258 c.806G>A p.R269H Chr.2: 232457817-232457817 on Build GRCh38
3049257 c.805C>T p.R269C Chr.2: 232457816-232457816 on Build GRCh38
1692166 c.730G>A p.G244R Chr.2: 232457646-232457646 on Build GRCh38
3909833 c.737G>A p.R246K Chr.2: 232457653-232457653 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547308328 Chr.2: 232457730-232457730 on Build GRCh38
rs761405805 Chr.2: 232457799-232457799 on Build GRCh38
rs1025185454 Chr.2: 232457622-232457622 on Build GRCh38
rs772086633 Chr.2: 232457711-232457711 on Build GRCh38
rs779212935 Chr.2: 232457747-232457747 on Build GRCh38
rs764771831 Chr.2: 232457674-232457674 on Build GRCh38
rs141754817 Chr.2: 232457660-232457660 on Build GRCh38
rs777784785 Chr.2: 232457756-232457756 on Build GRCh38
rs114417218 Chr.2: 232457707-232457707 on Build GRCh38
rs780997860 Chr.2: 232457689-232457689 on Build GRCh38
rs144606029 Chr.2: 232457606-232457606 on Build GRCh38
rs371346221 Chr.2: 232457626-232457626 on Build GRCh38
rs746297760 Chr.2: 232457772-232457772 on Build GRCh38
rs751576029 Chr.2: 232457734-232457734 on Build GRCh38
rs114286057 Chr.2: 232457729-232457729 on Build GRCh38
rs769995763 Chr.2: 232457775-232457775 on Build GRCh38
rs755847941 Chr.2: 232457740-232457740 on Build GRCh38
rs931905027 Chr.2: 232457741-232457741 on Build GRCh38
rs774541410 Chr.2: 232457615-232457615 on Build GRCh38
rs751483451 Chr.2: 232457680-232457680 on Build GRCh38
rs775212105 Chr.2: 232457779-232457779 on Build GRCh38
rs372337112 Chr.2: 232457726-232457726 on Build GRCh38
rs771505509 Chr.2: 232457769-232457769 on Build GRCh38
rs199684542 Chr.2: 232457748-232457748 on Build GRCh38
rs766666691 Chr.2: 232457661-232457661 on Build GRCh38
rs1014011715 Chr.2: 232457621-232457621 on Build GRCh38
rs777100178 Chr.2: 232457718-232457718 on Build GRCh38
rs773699984 Chr.2: 232457645-232457645 on Build GRCh38
rs781543646 Chr.2: 232457771-232457771 on Build GRCh38
rs748962802 Chr.2: 232457698-232457699 on Build GRCh38
rs761893711 Chr.2: 232457735-232457735 on Build GRCh38
rs757100455 Chr.2: 232457600-232457600 on Build GRCh38
rs752287804 Chr.2: 232457676-232457676 on Build GRCh38
rs778998833 Chr.2: 232457703-232457703 on Build GRCh38
rs202041605 Chr.2: 232457719-232457719 on Build GRCh38
rs386656273 Chr.2: 232457714-232457715 on Build GRCh38
rs762653548 Chr.2: 232457781-232457781 on Build GRCh38
rs747066349 Chr.2: 232457761-232457761 on Build GRCh38
rs772633773 Chr.2: 232457642-232457642 on Build GRCh38
rs1050394651 Chr.2: 232457762-232457762 on Build GRCh38
rs374789561 Chr.2: 232457696-232457696 on Build GRCh38
rs748146535 Chr.2: 232457634-232457634 on Build GRCh38
rs768302710 Chr.2: 232457788-232457788 on Build GRCh38
rs2246004 Chr.2: 232457714-232457714 on Build GRCh38
rs774047394 Chr.2: 232457796-232457796 on Build GRCh38
rs757973136 Chr.2: 232457679-232457679 on Build GRCh38
rs757120743 Chr.2: 232457688-232457688 on Build GRCh38
rs758508313 Chr.2: 232457750-232457750 on Build GRCh38
rs6708439 Chr.2: 232457715-232457715 on Build GRCh38
rs763630188 Chr.2: 232457732-232457732 on Build GRCh38
rs143617869 Chr.2: 232457612-232457612 on Build GRCh38
rs759176560 Chr.2: 232457670-232457670 on Build GRCh38
rs866432794 Chr.2: 232457708-232457708 on Build GRCh38
rs780434324 Chr.2: 232457605-232457605 on Build GRCh38
rs771310081 Chr.2: 232457716-232457716 on Build GRCh38
rs750299899 Chr.2: 232457739-232457739 on Build GRCh38

View Full Product Details