Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96840089-96840338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD23
Entrez Gene ID
200539
Gene Name
ankyrin repeat domain 23
Gene Aliases
DARP, MARP3
UniGene
Hs.643430
Gene Chromosome Location
Chr.2: 96837914-96844021 on Build GRCh38
Transcripts
NM_144994.7

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3764866 c.625A>T p.I209F Chr.2: 96840095-96840095 on Build GRCh38
5719633 c.598G>T p.G200* Chr.2: 96840258-96840258 on Build GRCh38
4759136 c.580A>C p.K194Q Chr.2: 96840276-96840276 on Build GRCh38
2825850 c.615C>T p.A205A Chr.2: 96840241-96840241 on Build GRCh38
6028412 c.605G>A p.R202Q Chr.2: 96840251-96840251 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762839774 Chr.2: 96840118-96840118 on Build GRCh38
rs772252975 Chr.2: 96840297-96840297 on Build GRCh38
rs750102963 Chr.2: 96840157-96840157 on Build GRCh38
rs924229163 Chr.2: 96840249-96840249 on Build GRCh38
rs760660425 Chr.2: 96840221-96840221 on Build GRCh38
rs939667883 Chr.2: 96840243-96840243 on Build GRCh38
rs748395148 Chr.2: 96840216-96840216 on Build GRCh38
rs776258812 Chr.2: 96840227-96840227 on Build GRCh38
rs374283622 Chr.2: 96840220-96840220 on Build GRCh38
rs752337725 Chr.2: 96840240-96840240 on Build GRCh38
rs756999496 Chr.2: 96840136-96840137 on Build GRCh38
rs757645814 Chr.2: 96840109-96840109 on Build GRCh38
rs746828314 Chr.2: 96840306-96840306 on Build GRCh38
rs750811588 Chr.2: 96840250-96840250 on Build GRCh38
rs375676500 Chr.2: 96840177-96840177 on Build GRCh38
rs778979801 Chr.2: 96840274-96840274 on Build GRCh38
rs766286335 Chr.2: 96840225-96840225 on Build GRCh38
rs774514317 Chr.2: 96840212-96840212 on Build GRCh38
rs990496445 Chr.2: 96840191-96840191 on Build GRCh38
rs756645429 Chr.2: 96840183-96840183 on Build GRCh38
rs991788490 Chr.2: 96840139-96840139 on Build GRCh38
rs767919060 Chr.2: 96840248-96840248 on Build GRCh38
rs768832356 Chr.2: 96840206-96840206 on Build GRCh38
rs746319902 Chr.2: 96840140-96840140 on Build GRCh38
rs377168591 Chr.2: 96840314-96840314 on Build GRCh38
rs570484917 Chr.2: 96840251-96840251 on Build GRCh38
rs774109767 Chr.2: 96840201-96840201 on Build GRCh38
rs918119685 Chr.2: 96840203-96840203 on Build GRCh38
rs777614482 Chr.2: 96840298-96840298 on Build GRCh38
rs759158996 Chr.2: 96840231-96840231 on Build GRCh38
rs113201764 Chr.2: 96840254-96840254 on Build GRCh38
rs186341431 Chr.2: 96840108-96840108 on Build GRCh38
rs778695344 Chr.2: 96840304-96840304 on Build GRCh38
rs771844708 Chr.2: 96840218-96840218 on Build GRCh38
rs759558510 Chr.2: 96840317-96840317 on Build GRCh38
rs147978079 Chr.2: 96840284-96840284 on Build GRCh38
rs149251479 Chr.2: 96840239-96840239 on Build GRCh38
rs766373535 Chr.2: 96840155-96840155 on Build GRCh38
rs781421350 Chr.2: 96840138-96840138 on Build GRCh38
rs770871544 Chr.2: 96840313-96840313 on Build GRCh38
rs754370159 Chr.2: 96840259-96840259 on Build GRCh38
rs547188471 Chr.2: 96840167-96840167 on Build GRCh38
rs983846294 Chr.2: 96840179-96840179 on Build GRCh38
rs780165631 Chr.2: 96840193-96840193 on Build GRCh38
rs913315592 Chr.2: 96840236-96840237 on Build GRCh38
rs931490325 Chr.2: 96840172-96840172 on Build GRCh38
rs368416371 Chr.2: 96840252-96840252 on Build GRCh38
rs754999115 Chr.2: 96840262-96840262 on Build GRCh38

View Full Product Details