Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 173187443-173187700 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZAK
Entrez Gene ID
51776
Gene Name
sterile alpha motif and leucine zipper containing kinase AZK
Gene Aliases
AZK, MLK7, MLT, MLTK, MRK, SFMMP, mlklak, pk
UniGene
Hs.444451
Gene Chromosome Location
Chr.2: 173075435-173268009 on Build GRCh38
Transcripts
NM_016653.2 NM_133646.2 XM_005246640.2 XM_017004323.1 XM_017004324.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1009659 c.352A>C p.M118L Chr.2: 173187560-173187560 on Build GRCh38
4086815 c.377C>T p.P126L Chr.2: 173187585-173187585 on Build GRCh38
1009662 c.410G>T p.R137I Chr.2: 173187618-173187618 on Build GRCh38
1009661 c.410G>T p.R137I Chr.2: 173187618-173187618 on Build GRCh38
4086814 c.377C>T p.P126L Chr.2: 173187585-173187585 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747780250 Chr.2: 173187636-173187636 on Build GRCh38
rs754810552 Chr.2: 173187507-173187507 on Build GRCh38
rs769002914 Chr.2: 173187607-173187607 on Build GRCh38
rs1005520267 Chr.2: 173187572-173187572 on Build GRCh38
rs202049682 Chr.2: 173187622-173187622 on Build GRCh38
rs200359286 Chr.2: 173187504-173187504 on Build GRCh38
rs761269256 Chr.2: 173187599-173187599 on Build GRCh38
rs1048012738 Chr.2: 173187613-173187613 on Build GRCh38
rs750981685 Chr.2: 173187656-173187656 on Build GRCh38
rs775376620 Chr.2: 173187593-173187593 on Build GRCh38
rs775110895 Chr.2: 173187658-173187658 on Build GRCh38
rs762623276 Chr.2: 173187641-173187641 on Build GRCh38
rs767408002 Chr.2: 173187672-173187672 on Build GRCh38
rs769559063 Chr.2: 173187535-173187535 on Build GRCh38
rs781075192 Chr.2: 173187509-173187509 on Build GRCh38
rs371321751 Chr.2: 173187592-173187592 on Build GRCh38
rs773062350 Chr.2: 173187637-173187637 on Build GRCh38
rs747600196 Chr.2: 173187627-173187628 on Build GRCh38
rs779168226 Chr.2: 173187561-173187561 on Build GRCh38
rs765924759 Chr.2: 173187655-173187655 on Build GRCh38
rs534252285 Chr.2: 173187487-173187487 on Build GRCh38
rs754482945 Chr.2: 173187669-173187669 on Build GRCh38
rs922994929 Chr.2: 173187491-173187491 on Build GRCh38
rs746087074 Chr.2: 173187576-173187576 on Build GRCh38
rs113948503 Chr.2: 173187549-173187549 on Build GRCh38
rs200511415 Chr.2: 173187652-173187652 on Build GRCh38
rs762317045 Chr.2: 173187631-173187631 on Build GRCh38
rs558713642 Chr.2: 173187534-173187534 on Build GRCh38

View Full Product Details