Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35163858-35164087 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UNC45B
Entrez Gene ID
146862
Gene Name
unc-45 myosin chaperone B
Gene Aliases
CMYA4, CTRCT43, SMUNC45, UNC-45B, UNC45
UniGene
Hs.379636
Gene Chromosome Location
Chr.17: 35147793-35189345 on Build GRCh38
Transcripts
NM_001033576.1 NM_001267052.1 NM_001308281.1 NM_173167.3 XM_017024234.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6523442 c.1072C>A p.L358I Chr.17: 35164087-35164087 on Build GRCh38
6080583 c.1008G>T p.G336G Chr.17: 35164023-35164023 on Build GRCh38
4774485 c.1028C>A p.S343Y Chr.17: 35164043-35164043 on Build GRCh38
242177 c.1054C>G p.R352G Chr.17: 35164069-35164069 on Build GRCh38
4965200 c.1038C>G p.P346P Chr.17: 35164053-35164053 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs8079654 Chr.17: 35163972-35163972 on Build GRCh38
rs73288986 Chr.17: 35164050-35164050 on Build GRCh38
rs553976077 Chr.17: 35164014-35164014 on Build GRCh38
rs766093913 Chr.17: 35163958-35163958 on Build GRCh38
rs910831938 Chr.17: 35163927-35163927 on Build GRCh38
rs867174215 Chr.17: 35163942-35163942 on Build GRCh38
rs368480078 Chr.17: 35163897-35163897 on Build GRCh38
rs919444092 Chr.17: 35163891-35163891 on Build GRCh38
rs369464349 Chr.17: 35163974-35163974 on Build GRCh38
rs754299931 Chr.17: 35163968-35163968 on Build GRCh38
rs200248455 Chr.17: 35163955-35163955 on Build GRCh38
rs760206661 Chr.17: 35164056-35164056 on Build GRCh38
rs777783420 Chr.17: 35163985-35163985 on Build GRCh38
rs758516385 Chr.17: 35163984-35163984 on Build GRCh38
rs370514797 Chr.17: 35164036-35164036 on Build GRCh38
rs748091247 Chr.17: 35164045-35164045 on Build GRCh38
rs768722725 Chr.17: 35163977-35163977 on Build GRCh38
rs186108233 Chr.17: 35163921-35163921 on Build GRCh38
rs571014198 Chr.17: 35164051-35164051 on Build GRCh38
rs749115934 Chr.17: 35164026-35164026 on Build GRCh38
rs200767716 Chr.17: 35164035-35164035 on Build GRCh38
rs1048289026 Chr.17: 35164052-35164052 on Build GRCh38
rs140605574 Chr.17: 35164069-35164069 on Build GRCh38
rs1036860958 Chr.17: 35163976-35163976 on Build GRCh38
rs537852655 Chr.17: 35163893-35163893 on Build GRCh38
rs751629771 Chr.17: 35163999-35163999 on Build GRCh38
rs771429052 Chr.17: 35163947-35163947 on Build GRCh38
rs894425072 Chr.17: 35164059-35164059 on Build GRCh38
rs759929321 Chr.17: 35163971-35163971 on Build GRCh38
rs781343218 Chr.17: 35164023-35164023 on Build GRCh38

View Full Product Details