Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 85942219-85942450 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP24
Entrez Gene ID
83478
Gene Name
Rho GTPase activating protein 24
Gene Aliases
FILGAP, RC-GAP72, RCGAP72, p73, p73RhoGAP
UniGene
Hs.444229
Gene Chromosome Location
Chr.4: 85475131-86002670 on Build GRCh38
Transcripts
NM_001025616.2 NM_001042669.1 NM_001287805.1 NM_031305.2 XM_011532300.2 XM_017008679.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6469729 c.276G>T p.E92D Chr.4: 85942229-85942229 on Build GRCh38
6469728 c.555G>T p.E185D Chr.4: 85942229-85942229 on Build GRCh38
4640605 c.296G>A p.C99Y Chr.4: 85942249-85942249 on Build GRCh38
4126484 c.306G>T p.K102N Chr.4: 85942259-85942259 on Build GRCh38
3946613 c.309A>G p.P103P Chr.4: 85942262-85942262 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11097083 Chr.4: 85942314-85942314 on Build GRCh38
rs1051254165 Chr.4: 85942385-85942385 on Build GRCh38
rs377337392 Chr.4: 85942268-85942268 on Build GRCh38
rs1024194647 Chr.4: 85942336-85942336 on Build GRCh38
rs756591448 Chr.4: 85942306-85942306 on Build GRCh38
rs201176700 Chr.4: 85942285-85942285 on Build GRCh38
rs905784378 Chr.4: 85942382-85942382 on Build GRCh38
rs200070579 Chr.4: 85942303-85942303 on Build GRCh38
rs1033442070 Chr.4: 85942396-85942396 on Build GRCh38
rs768047025 Chr.4: 85942299-85942299 on Build GRCh38
rs369109146 Chr.4: 85942421-85942421 on Build GRCh38
rs930401408 Chr.4: 85942278-85942278 on Build GRCh38
rs764935952 Chr.4: 85942259-85942259 on Build GRCh38
rs1014173845 Chr.4: 85942316-85942316 on Build GRCh38
rs899877689 Chr.4: 85942350-85942350 on Build GRCh38
rs373851586 Chr.4: 85942262-85942262 on Build GRCh38
rs535525930 Chr.4: 85942331-85942331 on Build GRCh38
rs766486850 Chr.4: 85942313-85942313 on Build GRCh38
rs903387078 Chr.4: 85942386-85942386 on Build GRCh38
rs941313044 Chr.4: 85942315-85942315 on Build GRCh38
rs998276786 Chr.4: 85942367-85942367 on Build GRCh38
rs568792720 Chr.4: 85942383-85942383 on Build GRCh38

View Full Product Details