Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 66163813-66164048 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SREK1
Entrez Gene ID
140890
Gene Name
splicing regulatory glutamic acid and lysine rich protein 1
Gene Aliases
SFRS12, SRrp508, SRrp86
UniGene
Hs.49367
Gene Chromosome Location
Chr.5: 66144218-66183616 on Build GRCh38
Transcripts
NM_001077199.2 NM_001270492.1 NM_001323527.1 NM_001323529.1 NM_001323533.1 NM_001323534.1 NM_001323535.1 NM_139168.3 XM_011543171.2 XM_017009055.1 XM_017009056.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4950341 c.822G>C p.E274D Chr.5: 66163858-66163858 on Build GRCh38
6790171 c.472G>C p.E158Q Chr.5: 66163856-66163856 on Build GRCh38
4950342 c.474G>C p.E158D Chr.5: 66163858-66163858 on Build GRCh38
6790170 c.820G>C p.E274Q Chr.5: 66163856-66163856 on Build GRCh38
3617502 c.441C>T p.P147P Chr.5: 66163825-66163825 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750884844 Chr.5: 66163910-66163910 on Build GRCh38
rs747872574 Chr.5: 66163930-66163930 on Build GRCh38
rs763220438 Chr.5: 66163958-66163958 on Build GRCh38
rs1014956860 Chr.5: 66163934-66163934 on Build GRCh38
rs760611158 Chr.5: 66163974-66163974 on Build GRCh38
rs753309191 Chr.5: 66163882-66163882 on Build GRCh38
rs773020772 Chr.5: 66163967-66163967 on Build GRCh38
rs1009355069 Chr.5: 66163926-66163926 on Build GRCh38
rs756021816 Chr.5: 66163862-66163862 on Build GRCh38
rs994592640 Chr.5: 66163963-66163963 on Build GRCh38
rs958841701 Chr.5: 66164014-66164014 on Build GRCh38
rs779575332 Chr.5: 66163878-66163878 on Build GRCh38
rs374502706 Chr.5: 66163950-66163950 on Build GRCh38
rs183035989 Chr.5: 66163927-66163927 on Build GRCh38
rs754564316 Chr.5: 66163897-66163897 on Build GRCh38
rs769957784 Chr.5: 66163951-66163951 on Build GRCh38
rs750348070 Chr.5: 66163843-66163843 on Build GRCh38
rs3763061 Chr.5: 66163945-66163945 on Build GRCh38
rs543522109 Chr.5: 66163965-66163965 on Build GRCh38
rs3834832 Chr.5: 66163936-66163938 on Build GRCh38
rs1037012848 Chr.5: 66163962-66163962 on Build GRCh38
rs776727654 Chr.5: 66163947-66163947 on Build GRCh38
rs778292602 Chr.5: 66163944-66163944 on Build GRCh38
rs771375964 Chr.5: 66163946-66163946 on Build GRCh38
rs772548743 Chr.5: 66163936-66163936 on Build GRCh38
rs766763561 Chr.5: 66163963-66163963 on Build GRCh38
rs1027461638 Chr.5: 66163970-66163970 on Build GRCh38
rs369896734 Chr.5: 66163935-66163935 on Build GRCh38

View Full Product Details