Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109235023-109235268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SARS
Entrez Gene ID
6301
Gene Name
seryl-tRNA synthetase
Gene Aliases
SERRS, SERS
UniGene
Hs.531176
Gene Chromosome Location
Chr.1: 109213893-109238182 on Build GRCh38
Transcripts
NM_006513.3 XM_006710813.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3704805 c.801C>A p.Y267* Chr.1: 109235263-109235263 on Build GRCh38
7161665 c.773C>T p.S258F Chr.1: 109235235-109235235 on Build GRCh38
7150476 c.793G>A p.E265K Chr.1: 109235255-109235255 on Build GRCh38
247178 c.774T>C p.S258S Chr.1: 109235236-109235236 on Build GRCh38
7161666 c.773C>T p.S258F Chr.1: 109235235-109235235 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs55712540 Chr.1: 109235117-109235117 on Build GRCh38
rs954003659 Chr.1: 109235093-109235093 on Build GRCh38
rs918551311 Chr.1: 109235170-109235170 on Build GRCh38
rs771374380 Chr.1: 109235195-109235198 on Build GRCh38
rs375846713 Chr.1: 109235159-109235159 on Build GRCh38
rs777313469 Chr.1: 109235185-109235185 on Build GRCh38
rs527605218 Chr.1: 109235065-109235065 on Build GRCh38
rs372709506 Chr.1: 109235195-109235195 on Build GRCh38
rs201389158 Chr.1: 109235166-109235166 on Build GRCh38
rs753659098 Chr.1: 109235118-109235118 on Build GRCh38
rs114166723 Chr.1: 109235142-109235142 on Build GRCh38
rs939479076 Chr.1: 109235194-109235194 on Build GRCh38
rs959780959 Chr.1: 109235113-109235113 on Build GRCh38
rs542144392 Chr.1: 109235128-109235128 on Build GRCh38
rs529747277 Chr.1: 109235169-109235169 on Build GRCh38
rs576435150 Chr.1: 109235123-109235123 on Build GRCh38
rs563299382 Chr.1: 109235136-109235136 on Build GRCh38
rs757894839 Chr.1: 109235183-109235183 on Build GRCh38
rs370663371 Chr.1: 109235203-109235203 on Build GRCh38
rs368942213 Chr.1: 109235182-109235182 on Build GRCh38
rs775876959 Chr.1: 109235228-109235228 on Build GRCh38
rs925875863 Chr.1: 109235229-109235229 on Build GRCh38

View Full Product Details