Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49807901-49808146 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBA7
Entrez Gene ID
7318
Gene Name
ubiquitin like modifier activating enzyme 7
Gene Aliases
D8, UBA1B, UBE1L, UBE2
UniGene
Hs.16695
Gene Chromosome Location
Chr.3: 49805205-49813958 on Build GRCh38
Transcripts
NM_003335.2 XM_005265430.2 XM_006713321.2 XM_011534070.1 XM_011534071.1 XM_011534072.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7265396 c.2515C>T p.R839C Chr.3: 49808028-49808028 on Build GRCh38
7262322 c.2470G>A p.A824T Chr.3: 49808073-49808073 on Build GRCh38
446726 c.2445C>A p.N815K Chr.3: 49808098-49808098 on Build GRCh38
6098035 c.2520C>T p.A840A Chr.3: 49808023-49808023 on Build GRCh38
7324043 c.2511C>T p.V837V Chr.3: 49808032-49808032 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751934856 Chr.3: 49808028-49808028 on Build GRCh38
rs756147311 Chr.3: 49808039-49808039 on Build GRCh38
rs142428393 Chr.3: 49808035-49808035 on Build GRCh38
rs766945118 Chr.3: 49808115-49808115 on Build GRCh38
rs761411819 Chr.3: 49808113-49808113 on Build GRCh38
rs548053031 Chr.3: 49808077-49808077 on Build GRCh38
rs377294431 Chr.3: 49807964-49807964 on Build GRCh38
rs968084488 Chr.3: 49808066-49808066 on Build GRCh38
rs757122398 Chr.3: 49808063-49808063 on Build GRCh38
rs775226449 Chr.3: 49808001-49808001 on Build GRCh38
rs769276139 Chr.3: 49807973-49807973 on Build GRCh38
rs369854712 Chr.3: 49808047-49808047 on Build GRCh38
rs767612477 Chr.3: 49808036-49808036 on Build GRCh38
rs745611751 Chr.3: 49808068-49808068 on Build GRCh38
rs747459864 Chr.3: 49807945-49807945 on Build GRCh38
rs923590782 Chr.3: 49807979-49807979 on Build GRCh38
rs776146772 Chr.3: 49808094-49808094 on Build GRCh38
rs2234388 Chr.3: 49807998-49807998 on Build GRCh38
rs201100469 Chr.3: 49808038-49808038 on Build GRCh38
rs201812658 Chr.3: 49807986-49807986 on Build GRCh38
rs745670939 Chr.3: 49807990-49807990 on Build GRCh38
rs771669758 Chr.3: 49807991-49807991 on Build GRCh38
rs2234389 Chr.3: 49807936-49807936 on Build GRCh38
rs114272505 Chr.3: 49808010-49808010 on Build GRCh38
rs755192095 Chr.3: 49807922-49807922 on Build GRCh38
rs2230149 Chr.3: 49808093-49808093 on Build GRCh38
rs777971109 Chr.3: 49808046-49808046 on Build GRCh38
rs118020287 Chr.3: 49808027-49808027 on Build GRCh38
rs770703127 Chr.3: 49807976-49807976 on Build GRCh38
rs778850727 Chr.3: 49808067-49808067 on Build GRCh38
rs373286705 Chr.3: 49808075-49808075 on Build GRCh38
rs988606139 Chr.3: 49808107-49808107 on Build GRCh38
rs142820056 Chr.3: 49808049-49808049 on Build GRCh38
rs756267156 Chr.3: 49807954-49807954 on Build GRCh38
rs556229954 Chr.3: 49808118-49808118 on Build GRCh38
rs886640991 Chr.3: 49808022-49808022 on Build GRCh38
rs760078681 Chr.3: 49808009-49808009 on Build GRCh38
rs777542027 Chr.3: 49807963-49807963 on Build GRCh38
rs549814453 Chr.3: 49807932-49807932 on Build GRCh38
rs771898882 Chr.3: 49808074-49808074 on Build GRCh38
rs200445172 Chr.3: 49808079-49808079 on Build GRCh38

View Full Product Details