Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 12702328-12702569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LURAP1L-AS1
Entrez Gene ID
101929467
Gene Name
LURAP1L antisense RNA 1
Gene Aliases
-
UniGene
Hs.586443
Gene Chromosome Location
Chr.9: 12699999-12814389 on Build GRCh38
Transcripts
NR_125775.1 NM_000550.2

Gene Symbol
TYRP1
Entrez Gene ID
7306
Gene Name
tyrosinase related protein 1
Gene Aliases
CAS2, CATB, GP75, OCA3, TRP, TRP1, TYRP, b-PROTEIN
UniGene
Hs.270279
Gene Chromosome Location
Chr.9: 12693385-12710266 on Build GRCh38
Transcripts
NR_125775.1 NM_000550.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1636022 c.1033C>A p.P345T Chr.9: 12702390-12702390 on Build GRCh38
5711141 c.1074_1076delAGT p.V359delV Chr.9: 12702431-12702433 on Build GRCh38
4171844 c.1043A>T p.Y348F Chr.9: 12702400-12702400 on Build GRCh38
166262 c.1054_1057delACAA p.N353fs*31 Chr.9: 12702411-12702414 on Build GRCh38
1105048 c.1037C>A p.P346H Chr.9: 12702394-12702394 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs949437643 Chr.9: 12702438-12702438 on Build GRCh38
rs281865424 Chr.9: 12702424-12702424 on Build GRCh38
rs745690767 Chr.9: 12702356-12702356 on Build GRCh38
rs1002065780 Chr.9: 12702468-12702468 on Build GRCh38
rs781198356 Chr.9: 12702445-12702445 on Build GRCh38
rs767629323 Chr.9: 12702384-12702384 on Build GRCh38
rs41306302 Chr.9: 12702469-12702469 on Build GRCh38
rs756666042 Chr.9: 12702432-12702432 on Build GRCh38
rs35517777 Chr.9: 12702399-12702399 on Build GRCh38
rs35642605 Chr.9: 12702463-12702463 on Build GRCh38
rs746135416 Chr.9: 12702354-12702354 on Build GRCh38
rs41302218 Chr.9: 12702484-12702484 on Build GRCh38
rs758093517 Chr.9: 12702423-12702423 on Build GRCh38
rs113146199 Chr.9: 12702385-12702385 on Build GRCh38
rs145061390 Chr.9: 12702386-12702386 on Build GRCh38
rs1038790420 Chr.9: 12702381-12702381 on Build GRCh38
rs41302216 Chr.9: 12702511-12702511 on Build GRCh38
rs765656070 Chr.9: 12702404-12702404 on Build GRCh38
rs754238345 Chr.9: 12702390-12702390 on Build GRCh38
rs201325712 Chr.9: 12702478-12702478 on Build GRCh38
rs570312162 Chr.9: 12702449-12702449 on Build GRCh38
rs369286365 Chr.9: 12702357-12702357 on Build GRCh38
rs766894672 Chr.9: 12702389-12702389 on Build GRCh38
rs777720589 Chr.9: 12702475-12702475 on Build GRCh38
rs373011697 Chr.9: 12702431-12702431 on Build GRCh38
rs200277477 Chr.9: 12702459-12702459 on Build GRCh38
rs41305296 Chr.9: 12702532-12702532 on Build GRCh38
rs369807922 Chr.9: 12702411-12702411 on Build GRCh38
rs377679582 Chr.9: 12702394-12702394 on Build GRCh38
rs141098505 Chr.9: 12702353-12702353 on Build GRCh38
rs748407483 Chr.9: 12702467-12702467 on Build GRCh38
rs761970281 Chr.9: 12702359-12702359 on Build GRCh38
rs371052314 Chr.9: 12702456-12702456 on Build GRCh38
rs997888933 Chr.9: 12702526-12702526 on Build GRCh38
rs542025703 Chr.9: 12702491-12702491 on Build GRCh38
rs758302704 Chr.9: 12702445-12702446 on Build GRCh38
rs1030561709 Chr.9: 12702391-12702391 on Build GRCh38
rs1009608355 Chr.9: 12702480-12702480 on Build GRCh38
rs752447935 Chr.9: 12702420-12702420 on Build GRCh38
rs387906562 Chr.9: 12702414-12702417 on Build GRCh38
rs373723306 Chr.9: 12702388-12702388 on Build GRCh38

View Full Product Details