Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 36805157-36805418 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TPGS2
Entrez Gene ID
25941
Gene Name
tubulin polyglutamylase complex subunit 2
Gene Aliases
C18orf10, HMFN0601, L17, PGs2
UniGene
Hs.436636
Gene Chromosome Location
Chr.18: 36777647-36829393 on Build GRCh38
Transcripts
NM_001271949.1 NM_001271950.1 NM_001271951.1 NM_001271952.1 NM_001271953.1 NM_001271954.1 NM_001271955.1 NM_001271956.1 NM_015476.3 XM_005258242.3 XM_011525917.2 XM_017025699.1 XM_017025700.1 XM_017025701.1 XM_017025702.1 XM_017025703.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1611225 c.382G>T p.A128S Chr.18: 36805374-36805374 on Build GRCh38
6148652 c.348C>T p.P116P Chr.18: 36805408-36805408 on Build GRCh38
6148653 c.348C>T p.P116P Chr.18: 36805408-36805408 on Build GRCh38
6806791 c.370G>A p.D124N Chr.18: 36805386-36805386 on Build GRCh38
1388620 c.338C>T p.P113L Chr.18: 36805418-36805418 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199592671 Chr.18: 36805334-36805334 on Build GRCh38
rs370433122 Chr.18: 36805367-36805367 on Build GRCh38
rs936940368 Chr.18: 36805263-36805263 on Build GRCh38
rs572398777 Chr.18: 36805287-36805287 on Build GRCh38
rs750403506 Chr.18: 36805370-36805370 on Build GRCh38
rs7234328 Chr.18: 36805386-36805386 on Build GRCh38
rs150488919 Chr.18: 36805317-36805317 on Build GRCh38
rs890722739 Chr.18: 36805199-36805199 on Build GRCh38
rs994626834 Chr.18: 36805312-36805312 on Build GRCh38
rs781687680 Chr.18: 36805398-36805398 on Build GRCh38
rs1046538653 Chr.18: 36805318-36805318 on Build GRCh38
rs562703180 Chr.18: 36805392-36805392 on Build GRCh38
rs1003346004 Chr.18: 36805325-36805325 on Build GRCh38
rs755065481 Chr.18: 36805385-36805385 on Build GRCh38
rs895327010 Chr.18: 36805304-36805304 on Build GRCh38
rs762565101 Chr.18: 36805360-36805360 on Build GRCh38
rs140428446 Chr.18: 36805207-36805207 on Build GRCh38
rs758677668 Chr.18: 36805387-36805387 on Build GRCh38
rs777922899 Chr.18: 36805389-36805389 on Build GRCh38
rs114281017 Chr.18: 36805396-36805396 on Build GRCh38
rs769316832 Chr.18: 36805332-36805332 on Build GRCh38
rs751359617 Chr.18: 36805362-36805362 on Build GRCh38
rs1034783536 Chr.18: 36805327-36805327 on Build GRCh38

View Full Product Details