Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 38460780-38461023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP23
Entrez Gene ID
57636
Gene Name
Rho GTPase activating protein 23
Gene Aliases
-
UniGene
Hs.374446
Gene Chromosome Location
Chr.17: 38419286-38512392 on Build GRCh38
Transcripts
NM_001199417.1 XM_006721991.1 XM_006721992.3 XM_006721993.3 XM_006721994.3 XM_011525073.1 XM_011525074.2 XM_011525076.2 XM_011525077.2 XM_017024902.1

SNP Detail

SNP ID Chromosome Location
rs11869478 Chr.17: 38460832-38460832 on Build GRCh38
rs746335985 Chr.17: 38460953-38460953 on Build GRCh38
rs566326401 Chr.17: 38460905-38460905 on Build GRCh38
rs914230348 Chr.17: 38460918-38460918 on Build GRCh38
rs936930436 Chr.17: 38460831-38460831 on Build GRCh38
rs1035981578 Chr.17: 38460816-38460816 on Build GRCh38
rs969554370 Chr.17: 38460896-38460896 on Build GRCh38
rs780851692 Chr.17: 38460949-38460949 on Build GRCh38
rs1039532996 Chr.17: 38461003-38461003 on Build GRCh38
rs866800948 Chr.17: 38460924-38460924 on Build GRCh38
rs1007916926 Chr.17: 38460981-38460981 on Build GRCh38
rs1038094353 Chr.17: 38460801-38460801 on Build GRCh38
rs535251416 Chr.17: 38460948-38460948 on Build GRCh38
rs751100910 Chr.17: 38460947-38460947 on Build GRCh38
rs926765508 Chr.17: 38460810-38460810 on Build GRCh38
rs895955264 Chr.17: 38460826-38460826 on Build GRCh38
rs771135797 Chr.17: 38460979-38460979 on Build GRCh38
rs578153829 Chr.17: 38460975-38460975 on Build GRCh38
rs979230887 Chr.17: 38460921-38460921 on Build GRCh38
rs748579886 Chr.17: 38460923-38460923 on Build GRCh38
rs988009912 Chr.17: 38460965-38460965 on Build GRCh38
rs756603063 Chr.17: 38460955-38460955 on Build GRCh38
rs934392453 Chr.17: 38460875-38460875 on Build GRCh38
rs1023363243 Chr.17: 38460877-38460877 on Build GRCh38
rs9898977 Chr.17: 38460966-38460966 on Build GRCh38
rs897853634 Chr.17: 38460805-38460805 on Build GRCh38
rs993748251 Chr.17: 38460811-38460811 on Build GRCh38
rs763716843 Chr.17: 38460935-38460935 on Build GRCh38

View Full Product Details