Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 63095880-63096131 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC22A24
Entrez Gene ID
283238
Gene Name
solute carrier family 22 member 24
Gene Aliases
NET46
UniGene
Hs.375139
Gene Chromosome Location
Chr.11: 63079940-63144822 on Build GRCh38
Transcripts
NM_001136506.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5005715 c.994C>T p.R332* Chr.11: 63096067-63096067 on Build GRCh38
1355518 c.1024C>T p.R342C Chr.11: 63096037-63096037 on Build GRCh38
1355517 c.1039C>T p.R347* Chr.11: 63096022-63096022 on Build GRCh38
4619965 c.994C>G p.R332G Chr.11: 63096067-63096067 on Build GRCh38
6014467 c.1016C>T p.S339F Chr.11: 63096045-63096045 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551224216 Chr.11: 63096093-63096093 on Build GRCh38
rs147369451 Chr.11: 63096020-63096020 on Build GRCh38
rs574508599 Chr.11: 63095997-63095997 on Build GRCh38
rs184268615 Chr.11: 63095960-63095960 on Build GRCh38
rs554854763 Chr.11: 63095996-63095996 on Build GRCh38
rs374808719 Chr.11: 63096098-63096098 on Build GRCh38
rs532105556 Chr.11: 63096021-63096021 on Build GRCh38
rs1036356059 Chr.11: 63095922-63095922 on Build GRCh38
rs775530409 Chr.11: 63095931-63095931 on Build GRCh38
rs189467728 Chr.11: 63096047-63096047 on Build GRCh38
rs552962380 Chr.11: 63095955-63095955 on Build GRCh38
rs7113279 Chr.11: 63096046-63096046 on Build GRCh38
rs371518525 Chr.11: 63096066-63096066 on Build GRCh38
rs769074675 Chr.11: 63095989-63095989 on Build GRCh38
rs923395793 Chr.11: 63096091-63096091 on Build GRCh38
rs559053081 Chr.11: 63096030-63096030 on Build GRCh38
rs774735623 Chr.11: 63096055-63096056 on Build GRCh38
rs1049080829 Chr.11: 63096041-63096041 on Build GRCh38
rs766714390 Chr.11: 63096008-63096008 on Build GRCh38
rs755232708 Chr.11: 63096036-63096036 on Build GRCh38
rs915170214 Chr.11: 63095947-63095947 on Build GRCh38
rs750714307 Chr.11: 63095965-63095965 on Build GRCh38
rs142762522 Chr.11: 63095933-63095933 on Build GRCh38
rs370535827 Chr.11: 63096054-63096054 on Build GRCh38
rs528111522 Chr.11: 63096037-63096037 on Build GRCh38
rs997546849 Chr.11: 63095951-63095951 on Build GRCh38
rs957111531 Chr.11: 63095994-63095994 on Build GRCh38
rs763976918 Chr.11: 63096050-63096050 on Build GRCh38
rs756403957 Chr.11: 63095974-63095974 on Build GRCh38
rs376030248 Chr.11: 63096103-63096103 on Build GRCh38
rs543256896 Chr.11: 63096007-63096007 on Build GRCh38
rs553434553 Chr.11: 63095924-63095924 on Build GRCh38
rs780297849 Chr.11: 63095968-63095968 on Build GRCh38
rs374095536 Chr.11: 63096022-63096022 on Build GRCh38
rs754245809 Chr.11: 63096031-63096031 on Build GRCh38
rs1020034130 Chr.11: 63096005-63096005 on Build GRCh38
rs1050943376 Chr.11: 63095952-63095952 on Build GRCh38

View Full Product Details