Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 131587787-131588054 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAPGEF1
Entrez Gene ID
2889
Gene Name
Rap guanine nucleotide exchange factor 1
Gene Aliases
C3G, GRF2
UniGene
Hs.127897
Gene Chromosome Location
Chr.9: 131576770-131740068 on Build GRCh38
Transcripts
NM_001304275.1 NM_005312.3 NM_198679.1 XM_005272186.4 XM_005272191.3 XM_006717067.3 XM_006717072.3 XM_006717074.3 XM_011518569.2 XM_011518570.2 XM_011518571.2 XM_011518572.2 XM_011518573.2 XM_011518574.2 XM_011518575.2 XM_011518576.2 XM_011518577.2 XM_011518578.2 XM_011518579.2 XM_017014633.1 XM_017014634.1 XM_017014635.1 XM_017014636.1 XM_017014637.1 XM_017014638.1 XM_017014639.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4869199 c.847C>A p.L283I Chr.9: 131587957-131587957 on Build GRCh38
5743365 c.2612C>T p.A871V Chr.9: 131587964-131587964 on Build GRCh38
5984411 c.2581G>A p.E861K Chr.9: 131587995-131587995 on Build GRCh38
6305386 c.2638G>T p.E880* Chr.9: 131587824-131587824 on Build GRCh38
5984409 c.2671G>A p.E891K Chr.9: 131587794-131587794 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758856253 Chr.9: 131587867-131587867 on Build GRCh38
rs865972904 Chr.9: 131588018-131588018 on Build GRCh38
rs68140797 Chr.9: 131587903-131587906 on Build GRCh38
rs369423823 Chr.9: 131588024-131588024 on Build GRCh38
rs926957660 Chr.9: 131587996-131587996 on Build GRCh38
rs546298702 Chr.9: 131587899-131587899 on Build GRCh38
rs367627947 Chr.9: 131588020-131588020 on Build GRCh38
rs962237348 Chr.9: 131587880-131587880 on Build GRCh38
rs75325491 Chr.9: 131588003-131588003 on Build GRCh38
rs771884949 Chr.9: 131587827-131587827 on Build GRCh38
rs374141379 Chr.9: 131587860-131587860 on Build GRCh38
rs913955174 Chr.9: 131587818-131587818 on Build GRCh38
rs776370107 Chr.9: 131587851-131587851 on Build GRCh38
rs368356509 Chr.9: 131587894-131587894 on Build GRCh38
rs781276413 Chr.9: 131587896-131587896 on Build GRCh38
rs780376046 Chr.9: 131587904-131587904 on Build GRCh38
rs764582747 Chr.9: 131587868-131587868 on Build GRCh38
rs67451016 Chr.9: 131587906-131587909 on Build GRCh38
rs572619546 Chr.9: 131587853-131587853 on Build GRCh38
rs776310031 Chr.9: 131587885-131587885 on Build GRCh38
rs564703743 Chr.9: 131587923-131587923 on Build GRCh38
rs752065497 Chr.9: 131587876-131587876 on Build GRCh38
rs555493189 Chr.9: 131587813-131587813 on Build GRCh38
rs1014157185 Chr.9: 131587884-131587884 on Build GRCh38
rs1015013739 Chr.9: 131587856-131587856 on Build GRCh38
rs376086696 Chr.9: 131588023-131588023 on Build GRCh38
rs186725842 Chr.9: 131587987-131587987 on Build GRCh38
rs759986720 Chr.9: 131587909-131587910 on Build GRCh38
rs746961500 Chr.9: 131587937-131587937 on Build GRCh38
rs375486224 Chr.9: 131587975-131587975 on Build GRCh38
rs1055367249 Chr.9: 131587998-131587998 on Build GRCh38
rs760193016 Chr.9: 131587834-131587834 on Build GRCh38
rs756261682 Chr.9: 131587900-131587900 on Build GRCh38
rs773346882 Chr.9: 131587829-131587829 on Build GRCh38
rs771994429 Chr.9: 131587928-131587928 on Build GRCh38
rs372121770 Chr.9: 131587918-131587918 on Build GRCh38
rs773221718 Chr.9: 131587929-131587929 on Build GRCh38
rs766663248 Chr.9: 131587905-131587908 on Build GRCh38
rs765996716 Chr.9: 131587836-131587836 on Build GRCh38
rs768003752 Chr.9: 131587902-131587903 on Build GRCh38
rs764782256 Chr.9: 131588019-131588019 on Build GRCh38
rs770958816 Chr.9: 131587877-131587878 on Build GRCh38

View Full Product Details