Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 31259949-31260222 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNRNP40
Entrez Gene ID
9410
Gene Name
small nuclear ribonucleoprotein U5 subunit 40
Gene Aliases
40K, HPRP8BP, PRP8BP, PRPF8BP, SPF38, WDR57
UniGene
Hs.33962
Gene Chromosome Location
Chr.1: 31259568-31296797 on Build GRCh38
Transcripts
NM_004814.2

SNP Detail

SNP ID Chromosome Location
rs757534721 Chr.1: 31260018-31260018 on Build GRCh38
rs147710333 Chr.1: 31260067-31260067 on Build GRCh38
rs373867939 Chr.1: 31260026-31260026 on Build GRCh38
rs769710094 Chr.1: 31260029-31260029 on Build GRCh38
rs142558035 Chr.1: 31260190-31260190 on Build GRCh38
rs776177722 Chr.1: 31260123-31260123 on Build GRCh38
rs761331571 Chr.1: 31260065-31260065 on Build GRCh38
rs762244044 Chr.1: 31260159-31260175 on Build GRCh38
rs936916126 Chr.1: 31260081-31260081 on Build GRCh38
rs372944854 Chr.1: 31260141-31260141 on Build GRCh38
rs771494471 Chr.1: 31260059-31260059 on Build GRCh38
rs527421488 Chr.1: 31260066-31260066 on Build GRCh38
rs982285084 Chr.1: 31260054-31260054 on Build GRCh38
rs373342461 Chr.1: 31260044-31260044 on Build GRCh38
rs745563410 Chr.1: 31260160-31260160 on Build GRCh38
rs375822848 Chr.1: 31260119-31260119 on Build GRCh38
rs760235811 Chr.1: 31260168-31260168 on Build GRCh38
rs768393390 Chr.1: 31260108-31260108 on Build GRCh38
rs1041643519 Chr.1: 31260124-31260124 on Build GRCh38
rs776856044 Chr.1: 31260063-31260064 on Build GRCh38
rs928366723 Chr.1: 31260057-31260057 on Build GRCh38
rs750616795 Chr.1: 31260113-31260113 on Build GRCh38
rs926791558 Chr.1: 31260071-31260071 on Build GRCh38
rs199960563 Chr.1: 31260138-31260138 on Build GRCh38
rs2292165 Chr.1: 31260045-31260045 on Build GRCh38
rs372524542 Chr.1: 31260111-31260111 on Build GRCh38
rs972126644 Chr.1: 31259987-31259987 on Build GRCh38
rs962358799 Chr.1: 31259976-31259976 on Build GRCh38
rs1054449204 Chr.1: 31260102-31260102 on Build GRCh38
rs778976320 Chr.1: 31260019-31260019 on Build GRCh38
rs771750591 Chr.1: 31260162-31260162 on Build GRCh38
rs754299856 Chr.1: 31260069-31260069 on Build GRCh38
rs191285327 Chr.1: 31260120-31260120 on Build GRCh38
rs1055828812 Chr.1: 31260076-31260076 on Build GRCh38
rs754250681 Chr.1: 31260004-31260004 on Build GRCh38
rs764609959 Chr.1: 31259998-31259998 on Build GRCh38
rs371278262 Chr.1: 31260143-31260143 on Build GRCh38
rs770664729 Chr.1: 31260144-31260144 on Build GRCh38
rs368872878 Chr.1: 31260060-31260060 on Build GRCh38
rs778709111 Chr.1: 31260147-31260147 on Build GRCh38
rs1028087633 Chr.1: 31260178-31260178 on Build GRCh38
rs758436152 Chr.1: 31260020-31260020 on Build GRCh38
rs775180303 Chr.1: 31260166-31260166 on Build GRCh38
rs970851311 Chr.1: 31260050-31260050 on Build GRCh38
rs769298499 Chr.1: 31260139-31260139 on Build GRCh38
rs759190616 Chr.1: 31260051-31260051 on Build GRCh38
rs777617113 Chr.1: 31260043-31260043 on Build GRCh38
rs751637797 Chr.1: 31260133-31260133 on Build GRCh38

View Full Product Details