Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45178074-45178333 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAPPC6A
Entrez Gene ID
79090
Gene Name
trafficking protein particle complex 6A
Gene Aliases
TRS33
UniGene
Hs.466929
Gene Chromosome Location
Chr.19: 45162928-45178302 on Build GRCh38
Transcripts
NM_001270891.1 NM_001270892.1 NM_001270893.1 NM_024108.2 XM_017027295.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5726598 c.84G>A p.G28G Chr.19: 45178135-45178135 on Build GRCh38
5891132 c.100C>T p.R34C Chr.19: 45178119-45178119 on Build GRCh38
1394557 c.64G>T p.D22Y Chr.19: 45178155-45178155 on Build GRCh38
4515153 c.99C>T p.L33L Chr.19: 45178120-45178120 on Build GRCh38
7101893 c.52C>T p.L18L Chr.19: 45178167-45178167 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1054791528 Chr.19: 45178271-45178271 on Build GRCh38
rs759274260 Chr.19: 45178139-45178139 on Build GRCh38
rs1006427796 Chr.19: 45178312-45178312 on Build GRCh38
rs759219169 Chr.19: 45178206-45178206 on Build GRCh38
rs760258688 Chr.19: 45178249-45178249 on Build GRCh38
rs753077438 Chr.19: 45178294-45178294 on Build GRCh38
rs768657701 Chr.19: 45178179-45178179 on Build GRCh38
rs372125874 Chr.19: 45178122-45178122 on Build GRCh38
rs749167492 Chr.19: 45178239-45178239 on Build GRCh38
rs758737781 Chr.19: 45178229-45178229 on Build GRCh38
rs200256682 Chr.19: 45178258-45178258 on Build GRCh38
rs773273583 Chr.19: 45178184-45178184 on Build GRCh38
rs1043348870 Chr.19: 45178211-45178211 on Build GRCh38
rs762810119 Chr.19: 45178126-45178126 on Build GRCh38
rs374599713 Chr.19: 45178234-45178234 on Build GRCh38
rs575182789 Chr.19: 45178217-45178217 on Build GRCh38
rs749849940 Chr.19: 45178253-45178253 on Build GRCh38
rs896245968 Chr.19: 45178279-45178279 on Build GRCh38
rs775199670 Chr.19: 45178245-45178245 on Build GRCh38
rs978171451 Chr.19: 45178105-45178105 on Build GRCh38
rs557457387 Chr.19: 45178241-45178241 on Build GRCh38
rs796520413 Chr.19: 45178182-45178182 on Build GRCh38
rs772082934 Chr.19: 45178103-45178103 on Build GRCh38
rs1026454011 Chr.19: 45178286-45178286 on Build GRCh38
rs989408051 Chr.19: 45178170-45178170 on Build GRCh38
rs7254936 Chr.19: 45178302-45178302 on Build GRCh38
rs760231205 Chr.19: 45178218-45178218 on Build GRCh38
rs746025746 Chr.19: 45178153-45178153 on Build GRCh38
rs1026940477 Chr.19: 45178262-45178262 on Build GRCh38
rs373452379 Chr.19: 45178223-45178223 on Build GRCh38
rs750063749 Chr.19: 45178307-45178307 on Build GRCh38
rs778316421 Chr.19: 45178147-45178147 on Build GRCh38
rs577658969 Chr.19: 45178244-45178244 on Build GRCh38
rs150157536 Chr.19: 45178174-45178174 on Build GRCh38
rs140842439 Chr.19: 45178140-45178140 on Build GRCh38
rs745647720 Chr.19: 45178243-45178243 on Build GRCh38
rs373290714 Chr.19: 45178137-45178137 on Build GRCh38
rs1025079269 Chr.19: 45178093-45178093 on Build GRCh38
rs761433850 Chr.19: 45178197-45178198 on Build GRCh38
rs755067997 Chr.19: 45178233-45178233 on Build GRCh38
rs746146904 Chr.19: 45178101-45178101 on Build GRCh38
rs772371999 Chr.19: 45178160-45178160 on Build GRCh38
rs774289224 Chr.19: 45178204-45178204 on Build GRCh38
rs761613560 Chr.19: 45178111-45178111 on Build GRCh38
rs763570669 Chr.19: 45178221-45178221 on Build GRCh38
rs371151823 Chr.19: 45178251-45178251 on Build GRCh38
rs763037966 Chr.19: 45178188-45178188 on Build GRCh38
rs765547159 Chr.19: 45178224-45178224 on Build GRCh38
rs370815326 Chr.19: 45178232-45178232 on Build GRCh38
rs766340151 Chr.19: 45178134-45178134 on Build GRCh38
rs895036287 Chr.19: 45178270-45178270 on Build GRCh38
rs368881012 Chr.19: 45178209-45178209 on Build GRCh38
rs751256791 Chr.19: 45178156-45178167 on Build GRCh38
rs932209228 Chr.19: 45178205-45178205 on Build GRCh38
rs769790479 Chr.19: 45178118-45178118 on Build GRCh38
rs776150475 Chr.19: 45178250-45178250 on Build GRCh38
rs758784552 Chr.19: 45178265-45178265 on Build GRCh38
rs755631122 Chr.19: 45178143-45178143 on Build GRCh38
rs747087313 Chr.19: 45178177-45178177 on Build GRCh38
rs377732618 Chr.19: 45178231-45178231 on Build GRCh38
rs779264118 Chr.19: 45178150-45178150 on Build GRCh38
rs779948296 Chr.19: 45178166-45178166 on Build GRCh38
rs775645770 Chr.19: 45178106-45178106 on Build GRCh38
rs374135350 Chr.19: 45178230-45178230 on Build GRCh38
rs756830321 Chr.19: 45178196-45178196 on Build GRCh38
rs34006212 Chr.19: 45178136-45178136 on Build GRCh38
rs375637170 Chr.19: 45178195-45178195 on Build GRCh38
rs1052347100 Chr.19: 45178210-45178210 on Build GRCh38
rs377546640 Chr.19: 45178149-45178149 on Build GRCh38
rs779104175 Chr.19: 45178098-45178098 on Build GRCh38
rs752277364 Chr.19: 45178142-45178142 on Build GRCh38

View Full Product Details