Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 71072022-71072291 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZRANB2
Entrez Gene ID
9406
Gene Name
zinc finger RANBP2-type containing 2
Gene Aliases
ZIS, ZIS1, ZIS2, ZNF265
UniGene
Hs.194718
Gene Chromosome Location
Chr.1: 71063291-71081289 on Build GRCh38
Transcripts
NM_005455.4 NM_203350.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2241163 c.513+2T>C p.? Chr.1: 71072119-71072119 on Build GRCh38
2241175 c.383G>A p.G128E Chr.1: 71072251-71072251 on Build GRCh38
5902530 c.513+3G>A p.? Chr.1: 71072118-71072118 on Build GRCh38
4454074 c.399A>T p.K133N Chr.1: 71072235-71072235 on Build GRCh38
6009323 c.405A>G p.R135R Chr.1: 71072229-71072229 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774301600 Chr.1: 71072096-71072097 on Build GRCh38
rs1044840272 Chr.1: 71072166-71072166 on Build GRCh38
rs924179427 Chr.1: 71072060-71072060 on Build GRCh38
rs74090250 Chr.1: 71072066-71072066 on Build GRCh38
rs770182383 Chr.1: 71072244-71072244 on Build GRCh38
rs368681398 Chr.1: 71072095-71072095 on Build GRCh38
rs766215469 Chr.1: 71072124-71072124 on Build GRCh38
rs775699313 Chr.1: 71072245-71072245 on Build GRCh38
rs776941208 Chr.1: 71072217-71072217 on Build GRCh38
rs771308053 Chr.1: 71072213-71072213 on Build GRCh38
rs539994660 Chr.1: 71072051-71072051 on Build GRCh38
rs747768067 Chr.1: 71072113-71072113 on Build GRCh38
rs765390105 Chr.1: 71072107-71072107 on Build GRCh38
rs748668944 Chr.1: 71072094-71072094 on Build GRCh38
rs771220280 Chr.1: 71072100-71072100 on Build GRCh38
rs374317749 Chr.1: 71072232-71072232 on Build GRCh38
rs747865012 Chr.1: 71072085-71072085 on Build GRCh38
rs11209745 Chr.1: 71072260-71072260 on Build GRCh38
rs771877985 Chr.1: 71072222-71072222 on Build GRCh38
rs750307644 Chr.1: 71072117-71072117 on Build GRCh38
rs572969460 Chr.1: 71072106-71072106 on Build GRCh38
rs747588310 Chr.1: 71072177-71072177 on Build GRCh38
rs150313731 Chr.1: 71072181-71072181 on Build GRCh38
rs373990505 Chr.1: 71072072-71072072 on Build GRCh38
rs1021433786 Chr.1: 71072145-71072145 on Build GRCh38
rs561911391 Chr.1: 71072109-71072109 on Build GRCh38
rs754525484 Chr.1: 71072073-71072073 on Build GRCh38
rs759358285 Chr.1: 71072161-71072163 on Build GRCh38
rs772412017 Chr.1: 71072130-71072130 on Build GRCh38
rs778524864 Chr.1: 71072163-71072163 on Build GRCh38
rs141938859 Chr.1: 71072211-71072211 on Build GRCh38
rs777409783 Chr.1: 71072196-71072196 on Build GRCh38
rs753845132 Chr.1: 71072136-71072136 on Build GRCh38
rs541943177 Chr.1: 71072108-71072108 on Build GRCh38
rs754830965 Chr.1: 71072137-71072137 on Build GRCh38
rs966800257 Chr.1: 71072151-71072151 on Build GRCh38
rs778533279 Chr.1: 71072083-71072083 on Build GRCh38
rs747358620 Chr.1: 71072096-71072096 on Build GRCh38
rs777137518 Chr.1: 71072102-71072102 on Build GRCh38
rs778232335 Chr.1: 71072079-71072079 on Build GRCh38
rs900368635 Chr.1: 71072089-71072089 on Build GRCh38

View Full Product Details