Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 57688469-57688712 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZDHHC5
Entrez Gene ID
25921
Gene Name
zinc finger DHHC-type containing 5
Gene Aliases
DHHC5, ZNF375
UniGene
Hs.27239
Gene Chromosome Location
Chr.11: 57667836-57701187 on Build GRCh38
Transcripts
NM_015457.2 XM_011544899.1 XM_011544900.1 XM_011544901.1 XM_017017498.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6826171 c.337C>T p.R113C Chr.11: 57688618-57688618 on Build GRCh38
5583111 c.267C>T p.P89P Chr.11: 57688548-57688548 on Build GRCh38
7284674 c.293G>A p.G98D Chr.11: 57688574-57688574 on Build GRCh38
4197599 c.346C>T p.R116* Chr.11: 57688627-57688627 on Build GRCh38
4197596 c.305G>A p.R102H Chr.11: 57688586-57688586 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780731738 Chr.11: 57688598-57688598 on Build GRCh38
rs149824916 Chr.11: 57688572-57688572 on Build GRCh38
rs770126002 Chr.11: 57688674-57688674 on Build GRCh38
rs755790071 Chr.11: 57688600-57688600 on Build GRCh38
rs948535349 Chr.11: 57688662-57688662 on Build GRCh38
rs747499327 Chr.11: 57688633-57688633 on Build GRCh38
rs748385416 Chr.11: 57688672-57688672 on Build GRCh38
rs973737257 Chr.11: 57688562-57688562 on Build GRCh38
rs754471008 Chr.11: 57688597-57688597 on Build GRCh38
rs777308090 Chr.11: 57688668-57688668 on Build GRCh38
rs1041012361 Chr.11: 57688602-57688602 on Build GRCh38
rs773508584 Chr.11: 57688684-57688684 on Build GRCh38
rs776736903 Chr.11: 57688521-57688521 on Build GRCh38
rs377349008 Chr.11: 57688635-57688635 on Build GRCh38
rs139913783 Chr.11: 57688512-57688512 on Build GRCh38
rs746247101 Chr.11: 57688609-57688609 on Build GRCh38
rs780566939 Chr.11: 57688625-57688625 on Build GRCh38
rs762070478 Chr.11: 57688525-57688525 on Build GRCh38
rs932542538 Chr.11: 57688656-57688656 on Build GRCh38
rs772358812 Chr.11: 57688610-57688610 on Build GRCh38
rs751503725 Chr.11: 57688596-57688596 on Build GRCh38
rs201362018 Chr.11: 57688541-57688541 on Build GRCh38
rs201279144 Chr.11: 57688603-57688603 on Build GRCh38
rs752333870 Chr.11: 57688599-57688599 on Build GRCh38
rs143490442 Chr.11: 57688671-57688671 on Build GRCh38
rs763040057 Chr.11: 57688593-57688593 on Build GRCh38
rs766431909 Chr.11: 57688594-57688594 on Build GRCh38

View Full Product Details