Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 110028451-110028712 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD13A
Entrez Gene ID
88455
Gene Name
ankyrin repeat domain 13A
Gene Aliases
ANKRD13, NY-REN-25
UniGene
Hs.528703
Gene Chromosome Location
Chr.12: 109999169-110039432 on Build GRCh38
Transcripts
NM_033121.1 XM_005253980.2 XM_005253981.3 XM_005253982.3 XM_005253984.1 XM_011538937.1 XM_011538938.2 XM_017020157.1 XM_017020158.1 XM_017020159.1 XM_017020160.1 XM_017020161.1 XM_017020162.1 XM_017020163.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6331719 c.957G>A p.T319T Chr.12: 110028523-110028523 on Build GRCh38
6600366 c.1061C>T p.T354M Chr.12: 110028627-110028627 on Build GRCh38
4514952 c.992C>A p.T331K Chr.12: 110028558-110028558 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs769359726 Chr.12: 110028681-110028681 on Build GRCh38
rs780940221 Chr.12: 110028675-110028675 on Build GRCh38
rs762783172 Chr.12: 110028491-110028491 on Build GRCh38
rs553453027 Chr.12: 110028559-110028559 on Build GRCh38
rs926738902 Chr.12: 110028571-110028571 on Build GRCh38
rs367914311 Chr.12: 110028616-110028616 on Build GRCh38
rs769269891 Chr.12: 110028584-110028584 on Build GRCh38
rs370433255 Chr.12: 110028639-110028639 on Build GRCh38
rs764334581 Chr.12: 110028615-110028615 on Build GRCh38
rs113318525 Chr.12: 110028510-110028510 on Build GRCh38
rs146603421 Chr.12: 110028520-110028520 on Build GRCh38
rs917131043 Chr.12: 110028488-110028488 on Build GRCh38
rs768225979 Chr.12: 110028674-110028674 on Build GRCh38
rs886220092 Chr.12: 110028603-110028603 on Build GRCh38
rs758913072 Chr.12: 110028642-110028642 on Build GRCh38
rs766148522 Chr.12: 110028500-110028500 on Build GRCh38
rs776028800 Chr.12: 110028572-110028572 on Build GRCh38
rs757465378 Chr.12: 110028623-110028623 on Build GRCh38
rs771811225 Chr.12: 110028542-110028542 on Build GRCh38
rs760825839 Chr.12: 110028613-110028613 on Build GRCh38
rs747350769 Chr.12: 110028663-110028663 on Build GRCh38
rs762873412 Chr.12: 110028596-110028596 on Build GRCh38
rs949984719 Chr.12: 110028515-110028515 on Build GRCh38
rs751306654 Chr.12: 110028606-110028606 on Build GRCh38
rs755382995 Chr.12: 110028513-110028513 on Build GRCh38
rs150245310 Chr.12: 110028576-110028576 on Build GRCh38
rs753903377 Chr.12: 110028533-110028533 on Build GRCh38
rs756289642 Chr.12: 110028522-110028522 on Build GRCh38
rs779607468 Chr.12: 110028539-110028539 on Build GRCh38
rs780466148 Chr.12: 110028554-110028554 on Build GRCh38
rs534755664 Chr.12: 110028535-110028535 on Build GRCh38
rs779126124 Chr.12: 110028628-110028628 on Build GRCh38
rs766166768 Chr.12: 110028600-110028600 on Build GRCh38
rs746646331 Chr.12: 110028558-110028558 on Build GRCh38
rs141333386 Chr.12: 110028523-110028523 on Build GRCh38
rs747783874 Chr.12: 110028678-110028678 on Build GRCh38
rs141719454 Chr.12: 110028486-110028486 on Build GRCh38
rs1049280571 Chr.12: 110028588-110028588 on Build GRCh38
rs746358295 Chr.12: 110028549-110028549 on Build GRCh38
rs780295952 Chr.12: 110028650-110028650 on Build GRCh38
rs772626602 Chr.12: 110028552-110028552 on Build GRCh38
rs773190365 Chr.12: 110028590-110028590 on Build GRCh38

View Full Product Details