Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 78119095-78119338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG8
Entrez Gene ID
79053
Gene Name
ALG8, alpha-1,3-glucosyltransferase
Gene Aliases
CDG1H
UniGene
Hs.503368
Gene Chromosome Location
Chr.11: 78100942-78139660 on Build GRCh38
Transcripts
NM_001007027.2 NM_024079.4 XM_005274247.3 XM_011545252.2 XM_017018274.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6364718 c.482T>C p.I161T Chr.11: 78119246-78119246 on Build GRCh38
6256050 c.533C>A p.A178E Chr.11: 78119195-78119195 on Build GRCh38
6598472 c.487T>G p.F163V Chr.11: 78119241-78119241 on Build GRCh38
4661123 c.487T>G p.F163V Chr.11: 78119241-78119241 on Build GRCh38
4825117 c.484C>T p.H162Y Chr.11: 78119244-78119244 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375763823 Chr.11: 78119289-78119289 on Build GRCh38
rs779526652 Chr.11: 78119134-78119134 on Build GRCh38
rs758915269 Chr.11: 78119277-78119277 on Build GRCh38
rs369930977 Chr.11: 78119194-78119194 on Build GRCh38
rs777929727 Chr.11: 78119156-78119156 on Build GRCh38
rs746073749 Chr.11: 78119297-78119297 on Build GRCh38
rs771817128 Chr.11: 78119175-78119175 on Build GRCh38
rs373493408 Chr.11: 78119246-78119246 on Build GRCh38
rs369094234 Chr.11: 78119282-78119282 on Build GRCh38
rs773804345 Chr.11: 78119198-78119198 on Build GRCh38
rs759999973 Chr.11: 78119242-78119242 on Build GRCh38
rs753257659 Chr.11: 78119252-78119252 on Build GRCh38
rs577023142 Chr.11: 78119216-78119216 on Build GRCh38
rs777628869 Chr.11: 78119300-78119301 on Build GRCh38
rs751649169 Chr.11: 78119279-78119279 on Build GRCh38
rs750026741 Chr.11: 78119237-78119237 on Build GRCh38
rs757252497 Chr.11: 78119286-78119286 on Build GRCh38
rs372050285 Chr.11: 78119145-78119145 on Build GRCh38
rs769462985 Chr.11: 78119192-78119192 on Build GRCh38
rs375377321 Chr.11: 78119159-78119159 on Build GRCh38
rs762076956 Chr.11: 78119293-78119293 on Build GRCh38
rs778020413 Chr.11: 78119278-78119278 on Build GRCh38
rs745715068 Chr.11: 78119183-78119183 on Build GRCh38
rs146425128 Chr.11: 78119211-78119211 on Build GRCh38
rs200665495 Chr.11: 78119132-78119132 on Build GRCh38
rs748807034 Chr.11: 78119146-78119146 on Build GRCh38
rs755795309 Chr.11: 78119166-78119169 on Build GRCh38
rs186270173 Chr.11: 78119304-78119304 on Build GRCh38
rs761406814 Chr.11: 78119215-78119215 on Build GRCh38
rs762811727 Chr.11: 78119193-78119193 on Build GRCh38
rs978122618 Chr.11: 78119257-78119257 on Build GRCh38

View Full Product Details