Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 240575511-240575738 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNPEPL1
Entrez Gene ID
57140
Gene Name
arginyl aminopeptidase like 1
Gene Aliases
-
UniGene
Hs.5345
Gene Chromosome Location
Chr.2: 240568526-240578732 on Build GRCh38
Transcripts
NM_018226.4 XM_005247036.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1018728 c.769C>G p.L257V Chr.2: 240575562-240575562 on Build GRCh38
1264414 c.817G>A p.G273R Chr.2: 240575610-240575610 on Build GRCh38
5058783 c.801C>T p.S267S Chr.2: 240575594-240575594 on Build GRCh38
7387834 c.782C>T p.P261L Chr.2: 240575575-240575575 on Build GRCh38
1692252 c.803_804TG>AC p.V268D Chr.2: 240575596-240575597 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143454831 Chr.2: 240575674-240575674 on Build GRCh38
rs774548755 Chr.2: 240575549-240575549 on Build GRCh38
rs530188995 Chr.2: 240575682-240575682 on Build GRCh38
rs992834903 Chr.2: 240575646-240575646 on Build GRCh38
rs761914529 Chr.2: 240575659-240575659 on Build GRCh38
rs1031214603 Chr.2: 240575587-240575587 on Build GRCh38
rs774777756 Chr.2: 240575658-240575658 on Build GRCh38
rs866548119 Chr.2: 240575667-240575667 on Build GRCh38
rs892133209 Chr.2: 240575546-240575546 on Build GRCh38
rs762039090 Chr.2: 240575558-240575558 on Build GRCh38
rs767483184 Chr.2: 240575561-240575561 on Build GRCh38
rs989679540 Chr.2: 240575600-240575600 on Build GRCh38
rs769221485 Chr.2: 240575627-240575627 on Build GRCh38
rs754605463 Chr.2: 240575579-240575579 on Build GRCh38
rs147113035 Chr.2: 240575576-240575576 on Build GRCh38
rs778597551 Chr.2: 240575591-240575591 on Build GRCh38
rs890260581 Chr.2: 240575705-240575705 on Build GRCh38
rs376974000 Chr.2: 240575645-240575645 on Build GRCh38
rs138990862 Chr.2: 240575539-240575539 on Build GRCh38
rs931894764 Chr.2: 240575683-240575683 on Build GRCh38
rs372064131 Chr.2: 240575624-240575624 on Build GRCh38
rs755876911 Chr.2: 240575573-240575573 on Build GRCh38
rs749716938 Chr.2: 240575633-240575633 on Build GRCh38
rs780249265 Chr.2: 240575629-240575629 on Build GRCh38
rs907922142 Chr.2: 240575664-240575664 on Build GRCh38
rs766122737 Chr.2: 240575575-240575575 on Build GRCh38
rs918416568 Chr.2: 240575695-240575695 on Build GRCh38
rs539336061 Chr.2: 240575718-240575718 on Build GRCh38
rs1050699811 Chr.2: 240575701-240575701 on Build GRCh38
rs965586328 Chr.2: 240575552-240575552 on Build GRCh38
rs980694033 Chr.2: 240575596-240575596 on Build GRCh38
rs763298671 Chr.2: 240575534-240575534 on Build GRCh38
rs141589563 Chr.2: 240575595-240575595 on Build GRCh38
rs781682659 Chr.2: 240575604-240575604 on Build GRCh38
rs141937747 Chr.2: 240575594-240575594 on Build GRCh38
rs531855875 Chr.2: 240575567-240575567 on Build GRCh38

View Full Product Details