Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 230923748-230924021 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC13
Entrez Gene ID
79573
Gene Name
tetratricopeptide repeat domain 13
Gene Aliases
-
UniGene
Hs.424788
Gene Chromosome Location
Chr.1: 230906241-230978872 on Build GRCh38
Transcripts
NM_001122835.2 NM_024525.4 XM_005273260.1 XM_005273261.2 XM_005273262.1 XM_005273264.2 XM_006711814.2 XM_006711815.2 XM_011544275.2 XM_011544276.2 XM_017002321.1 XM_017002322.1 XM_017002323.1 XM_017002324.1 XM_017002325.1 XM_017002326.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6125405 c.1735G>T p.D579Y Chr.1: 230923920-230923920 on Build GRCh38
905405 c.1742C>A p.P581H Chr.1: 230923913-230923913 on Build GRCh38
5562692 c.1743C>G p.P581P Chr.1: 230923912-230923912 on Build GRCh38
3485040 c.1751G>C p.W584S Chr.1: 230923904-230923904 on Build GRCh38
5053195 c.1744G>A p.V582M Chr.1: 230923911-230923911 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748344205 Chr.1: 230923863-230923863 on Build GRCh38
rs750428126 Chr.1: 230923803-230923803 on Build GRCh38
rs760763450 Chr.1: 230923986-230923986 on Build GRCh38
rs749551835 Chr.1: 230923969-230923969 on Build GRCh38
rs544709293 Chr.1: 230923912-230923912 on Build GRCh38
rs770181698 Chr.1: 230923869-230923869 on Build GRCh38
rs763861787 Chr.1: 230923900-230923900 on Build GRCh38
rs1010196977 Chr.1: 230923913-230923913 on Build GRCh38
rs775897676 Chr.1: 230923802-230923804 on Build GRCh38
rs143159895 Chr.1: 230923855-230923855 on Build GRCh38
rs757069778 Chr.1: 230923795-230923795 on Build GRCh38
rs760068444 Chr.1: 230923895-230923895 on Build GRCh38
rs748510054 Chr.1: 230923992-230923992 on Build GRCh38
rs374496230 Chr.1: 230923959-230923959 on Build GRCh38
rs778691627 Chr.1: 230923794-230923806 on Build GRCh38
rs781215199 Chr.1: 230923952-230923952 on Build GRCh38
rs750126540 Chr.1: 230923917-230923917 on Build GRCh38
rs201348752 Chr.1: 230923883-230923883 on Build GRCh38
rs985138587 Chr.1: 230923788-230923788 on Build GRCh38
rs936871341 Chr.1: 230923824-230923824 on Build GRCh38
rs753717921 Chr.1: 230923793-230923793 on Build GRCh38
rs778127259 Chr.1: 230923968-230923968 on Build GRCh38
rs375078508 Chr.1: 230923804-230923804 on Build GRCh38
rs761527969 Chr.1: 230923909-230923909 on Build GRCh38
rs377133348 Chr.1: 230923858-230923858 on Build GRCh38
rs201925182 Chr.1: 230923924-230923924 on Build GRCh38
rs755095215 Chr.1: 230923949-230923949 on Build GRCh38
rs368389318 Chr.1: 230923871-230923871 on Build GRCh38
rs771288068 Chr.1: 230923884-230923884 on Build GRCh38
rs753594400 Chr.1: 230923908-230923908 on Build GRCh38
rs780194303 Chr.1: 230923820-230923820 on Build GRCh38
rs751678819 Chr.1: 230923948-230923948 on Build GRCh38
rs1041312262 Chr.1: 230923914-230923914 on Build GRCh38
rs41304010 Chr.1: 230923965-230923965 on Build GRCh38
rs779163475 Chr.1: 230923946-230923946 on Build GRCh38
rs1050198735 Chr.1: 230923780-230923780 on Build GRCh38
rs1025179209 Chr.1: 230923945-230923945 on Build GRCh38
rs746993584 Chr.1: 230923829-230923829 on Build GRCh38
rs771503454 Chr.1: 230923976-230923976 on Build GRCh38

View Full Product Details